Skip to Content
Merck
All Photos(3)

Key Documents

BMSE7500E

Benchmark QuadCount Automated Cell Counter

AC/DC input 230 V, EuroPlug

Synonym(s):

cell counter, hemocytometer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104000
NACRES:
NB.13

manufacturer/tradename

Benchmark Scientific E7500-E

Looking for similar products? Visit Product Comparison Guide

General description

The Accuris QuadCount improves on the simple technique of brightfield microscopy by utilizing an LED light source, precision optics and a high resolution CMOS detector. A finely tuned, motorized XYZ positioning stage provides for extra fine focusing and ability to capture multiple images. With count speeds at fast as 20 seconds per sample, the Accuris QuadCount provides many advantages over the tedious and time consuming manual hemocytometry method. The advanced software eliminates human bias in dead/live determinations, reduces counting errors, saves time, and improves efficiency.

Application

Automated cell counting

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service