Skip to Content
Merck
All Photos(1)

Key Documents

BAF203111718

SP Bel-Art® Safe-Lab Joint Tubing Adapter

for 24/40 Tapered Joints

Synonym(s):

adapter tubing

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122800
NACRES:
NB.43

Pricing and availability is not currently available.

packaging

pack of 1 ea

manufacturer/tradename

Bel-Art F20311-1718

hole diam.

10 mm

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
5499454995Z226688
L × O.D. × I.D.

10 ft × 1/16 in. × 0.010 in.

L × O.D. × I.D.

10 ft × 1/16 in. × 0.055 in.

L × O.D. × I.D.

10 ft × 1/8 in. × 0.062 in.

L × O.D. × I.D.

10 ft × 1/16 in. × 0.007 in.

color

blue

color

natural

color

natural

color

yellow

General description

Stopper has PTFE® PTFE body with Viton® O-ring.

Features and Benefits

  • Polypropylene screw cap on top has an inside O-ring for depth adjustment of glass tubing
  • Bottom inner joint has a threaded extracting nut for easy removal of adapter
  • Not Autoclavable


Legal Information

Bel-Art is a registered trademark of SP Scienceware, a division of SP
Teflon is a registered trademark of E. I. du Pont de Nemours and Company
Viton is a registered trademark of E. I. du Pont de Nemours and Company

  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Sorry, we don't have COAs for this product available online at this time.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service