Skip to Content
Merck
All Photos(4)

Key Documents

B78106

Sigma-Aldrich

1-Bromopropane

99%

Synonym(s):

Propyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2Br
CAS Number:
Molecular Weight:
122.99
Beilstein:
505936
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

4.3 (vs air)

vapor pressure

146 mmHg ( 20 °C)

Assay

99%

form

liquid

autoignition temp.

914 °F

refractive index

n20/D 1.434 (lit.)

bp

71 °C (lit.)

mp

−110 °C (lit.)

density

1.354 g/mL at 25 °C (lit.)

SMILES string

CCCBr

InChI

1S/C3H7Br/c1-2-3-4/h2-3H2,1H3

InChI key

CYNYIHKIEHGYOZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Bromopropane is used as a component of various models of binary mixtures, prepared to study their properties such as viscosity[1] and refractive index.[2]
It can be used as an alkylating reagent:
  • For the functionalization of seleno-substituted imidazo[1,2-α]pyridine derivatives.[3]
  • For the synthesis of 1,4-dihydroxy-2-propoxy anthraquinone derivatives.[4]

It can also be used as a reactant:
  • For the synthesis of Berberine-benzimidazole derivatives.[5]
  • For the synthesis of alkyl aryl ketones via carbonylative Barbier-Negishi coupling with aryl iodides.[6]

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 3 - Ozone 1 - Repr. 1B - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system, Liver,Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

76.1 °F - closed cup

Flash Point(C)

24.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Design, synthesis and biological evaluation of berberine-benzimidazole hybrids as new type of potentially DNA-targeting antimicrobial agents.
Jeyakkumar P, et al.
European Journal of Medicinal Chemistry, 122(11), 205-215 (2016)
Viscosity of pure organic liquids and binary liquid mixtures (2009)
Nickel-catalyzed C (sp2)-H selenation of imidazo [1, 2-?] pyridines with arylboronic acids or alkyl reagents using selenium powder.
Zhu J, et al.
Tetrahedron, 74(45), 6569-6576 (2018)
Sahabudeen Sheik Mohideen et al.
Toxicology, 285(1-2), 67-71 (2011-04-30)
1-Bromopropane (1-BP) has been used as an alternative to ozone-depleting solvents. Previous studies showed that 1-BP is neurotoxic in animals and humans. In humans, exposure to 1-BP caused various neurological and neurobehavioral symptoms or signs including depressive or irritated mood.
Makoto Samukawa et al.
Archives of internal medicine, 172(16), 1257-1260 (2012-08-16)
Health hazard alerts to 1-bromopropane, an alternative to ozone layer-damaging organic solvents, have been issued by some countries. Herein, we report a new case of 1-bromopropane-induced neurotoxicity. A 43-year-old male industrial worker developed muscle weakness, pain, numbness, and gait disturbance.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service