B45909
4,8-Bis(hydroxymethyl)tricyclo[5.2.1.02,6]decane, mixture of isomers
96%
Synonym(s):
TDMol, Tricyclo[5.2.1.02,6]decane-4,8-dimethanol
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Empirical Formula (Hill Notation):
C12H20O2
CAS Number:
Molecular Weight:
196.29
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
96%
form
liquid
refractive index
n20/D 1.528 (lit.)
SMILES string
OCC1CC2[C@H]3CC(CO)[C@H](C3)C2C1
InChI
1S/C12H20O2/c13-5-7-1-10-8-3-9(6-14)11(4-8)12(10)2-7/h7-14H,1-6H2/t7?,8-,9?,10?,11-,12?/m0/s1
InChI key
ZFZDWMXUMXACHS-IACGZSPGSA-N
Related Categories
Application
4,8-Bis(hydroxymethyl)tricyclo[5.2.1.02,6]decane (TDMol) is used in the preparation of high molecular weight poly(1,4-butylene carbonate) (PBC). It is also used in the synthesis of photoactive compounds (PAC) for the preparation of positive photo-resist materials.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
375.8 °F - closed cup
Flash Point(C)
191 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Preparation of macrodiols and polyols by chopping high?molecular?weight aliphatic polycarbonates.
Jeon J Y, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 52(11), 1570-1580 (2014)
Synthetic study and material applications for a positive photo resist of the acrylic series.
Cheng T S, et al.
Materials Letters, 57(3), 753-760 (2002)
Preparation for Acrylic Series Binders and Study of Thermal Property of Photo Resist.
Cheng T S
J. Polym. Mater., 27(4), 391-391 (2010)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service