Skip to Content
Merck
All Photos(1)

Key Documents

ALD00560

Sigma-Aldrich

1-((3,5-Difluorophenyl)sulfonyl)bicyclo[2.1.0]pentane

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H10F2O2S
Molecular Weight:
244.26
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

powder

reaction suitability

reaction type: C-C Bond Formation

storage temp.

2-8°C

SMILES string

O=S(C1=CC(F)=CC(F)=C1)(C23CCC2C3)=O

InChI

1S/C11H10F2O2S/c12-8-3-9(13)5-10(4-8)16(14,15)11-2-1-7(11)6-11/h3-5,7H,1-2,6H2

InChI key

IZBMPDKWKGIJQD-UHFFFAOYSA-N

Application

The following "spring-loaded" electrophile is a convenient reagent for the installation of cyclopentane to primary and secondary amines. This product is one of several synthetically useful reagents reported by Baran and coworkers on Strain-release amination.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Baran Group offers zinc-based reagents for versatile transformations in organic synthesis.

Baran Group offers zinc-based reagents for versatile transformations in organic synthesis.

Baran Group offers zinc-based reagents for versatile transformations in organic synthesis.

Baran Group offers zinc-based reagents for versatile transformations in organic synthesis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service