All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H11NaO2S
Molecular Weight:
170.21
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
form
powder
Quality Level
reaction suitability
reaction type: C-C Bond Formation
SMILES string
O=S(O[Na])C1CCCCC1
InChI
1S/C6H12O2S.Na/c7-9(8)6-4-2-1-3-5-6;/h6H,1-5H2,(H,7,8);/q;+1/p-1
InChI key
VKOSCBDFLPBMCB-UHFFFAOYSA-M
Related Categories
Application
The following Baran Sulfinate is a part of a toolbox of diversification reagents, which are ideal for the late-stage functionalization of nitrogen-containing heterocycles. The regioselectivity can be effectively tuned by modification of the pH and solvent selection.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service