All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C7H4FNO3S
Molecular Weight:
201.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
95% (GC)
form
solid
mp
36-41 °C
SMILES string
FS(OC1=CC=C(C#N)C=C1)(=O)=O
InChI
1S/C7H4FNO3S/c8-13(10,11)12-7-3-1-6(5-9)2-4-7/h1-4H
InChI key
QVLLNLVBRUJXHZ-UHFFFAOYSA-N
Application
The following aryl fluorosulfate can be utilized as a cross-coupling partner in Suzuki-Miyaura reactions. The standard reaction conditions for these palladium-catalyzed C-C bond formations are ligand-free and can be performed in water, at room temperature and are not air sensitive.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service