Skip to Content
Merck
All Photos(1)

Key Documents

935948

Sigma-Aldrich

Sodium hexafluorophosphate solution

new
greener alternative

in propylene carbonate, 1.0 M (NaPF6 in PC), battery grade

Synonym(s):

1.0 M NaPF6 in PC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352600

grade

battery grade

Quality Level

form

solution

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

0.95-1.05 M (NaPF6)
1.0 M (NaPF6 in PC)

impurities

≤100 ppm Acid
≤500.0 ppm H2O

anion traces

chloride (Cl-): ≤200 ppm
sulfate (SO42-): ≤200 ppm

application(s)

battery manufacturing

greener alternative category

General description

Sodium hexafluorophosphate solution in propylene carbonate (PC) is a 1.0 M solution of battery-grade NaPF6 dissolved in high-purity anhydrous propylene carbonate. The product is a clear solution with strictly minimized impurities including low moisture, low trace metals, low chloride, and low sulfate.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more information.

Application

1.0 M NaPF6 in propylene carbonate (PC), battery grade is a ready to use electrolyte for sodium-ion batteries., It is one of the best performing electrolytes, especially for hard carbon anodes. Propylene carbonate is an excellent solvent with a lower melting point (-49 °C) than other carbonates, making it suitable for low-temperature applications. For use with sodium metal anodes, much improved performance can be achieved by combining this electrolyte with 5 wt% fluoroethylene carbonate (FEC), which stabilizes the surface of the anode.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of advanced electrolytes in Na-ion batteries: application of the Red Moon method for molecular structure design of the SEI layer
Bouibes A, et al.
Royal Society of Chemistry, 12, 971-984 (2022)
High Stability and Long Cycle Life of Rechargeable Sodium-Ion Battery Using Manganese Oxide Cathode: A Combined Density Functional Theory (DFT) and Experimental Study
Pandit B, et al.
ACS Applied Materials & Interfaces, 11433-11441 (2021)
Electrolyte Solvation Structure Design for Sodium Ion Batteries
Tian Z, et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany) (2022)
Understanding the Electrochemical Compatibility and Reaction Mechanism on Na Metal and Hard Carbon Anodes of PC-Based Electrolytes for Sodium-Ion Batteries
Pan K, et al.
ACS Applied Materials & Interfaces, 10(46), 39651-39660 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service