Skip to Content
Merck
All Photos(2)

Key Documents

932140

Sigma-Aldrich

PXZ-DPS

≥97% (HPLC)

Synonym(s):

10,10′-(4,4′-Sulfonylbis(4,1-phenylene))bis(10H-phenoxazine)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H24N2O4S
CAS Number:
Molecular Weight:
580.65
MDL number:
UNSPSC Code:
12352005
NACRES:
NA.23
Assay:
≥97% (HPLC)
grade:
sublimed grade
solubility:
toluene: soluble

grade

sublimed grade

Quality Level

Assay

≥97% (HPLC)

energy gap

0.08 eV (delta EST)

description

2.8 eV (Eg: HOMO-LUMO gap)

loss

0.5% TGA, > 220 °C (weight loss)

solubility

toluene: soluble

fluorescence

λem 507 nm±10 nm (PL)

Orbital energy

HOMO - 5.59 eV 
LUMO - 2.79 eV 

Application

Dopant for Green TADF OLED, with a EQE of 17.5%.
PXZ-DPS can serve as an emitter or host material in OLEDs. Its unique molecular structure and photophysical properties contribute to efficient light emission and good device performance. It can be used as a donor material or incorporated in the active layer of organic photovoltaics (OPVs) devices. It can also be utilized in studies for the development of fluorescent chemosensors for detecting specific analytes or ions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficient blue organic light-emitting diodes employing thermally activated delayed fluorescence.
Zhang Q, et al.
Nature Photonics, 8, 326-332 (2014)
Recent progress of green thermally activated delayed fluorescent emitters
Im Y, et al.
Journal of information display, 18, 101-117 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service