928445
Droplet Generator Chip With Monitoring Chamber
Fluidic 1147, PC
Synonym(s):
Microfluidic chip, Polycarbonate
About This Item
Recommended Products
description
Microfludic chip x1
Looking for similar products? Visit Product Comparison Guide
Application
- Mini Luer Interface
- Material: Polycarbonate (PC)
- 4 droplet generator units
- Nozzle Dimensions: 70 µm and 80 µm
- 8 observation chambers
- Chamber Depth: 80 µm and 100 µm
- Chamber Volume: 6.8 µl and 8.5 µl
Features and Benefits
- All-in-one chip for droplet generation and observation
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Microfluidic assembly improves polyamine nanoencapsulation of nucleic acids, overcoming challenges like polydispersity and poor reproducibility.
Microfluidic assembly improves polyamine nanoencapsulation of nucleic acids, overcoming challenges like polydispersity and poor reproducibility.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service