Skip to Content
Merck
All Photos(1)

Documents

92768

Sigma-Aldrich

(Trimethylsilyl)methylamine

≥98.0% (GC)

Synonym(s):

(Aminomethyl)trimethylsilane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H13NSi
CAS Number:
Molecular Weight:
103.24
Beilstein:
1731488
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (GC)

form

liquid

refractive index

n20/D 1.416

bp

92-94 °C (lit.)

density

0.771 g/mL at 20 °C (lit.)

SMILES string

C[Si](C)(C)CN

InChI

1S/C4H13NSi/c1-6(2,3)4-5/h4-5H2,1-3H3

InChI key

YVWPNDBYAAEZBF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Synthesis of trimethylsilyl-diazomethane; 1,3-Cycloaddition of its imine; preparation of azomethine ylides from ketones

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

48.2 °F - closed cup

Flash Point(C)

9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

K. Achiwa et al.
Chemical & Pharmaceutical Bulletin, 31, 3939-3939 (1983)
D. Seyferth et al.
Journal of Organometallic Chemistry, 44, 279-279 (1972)
M. Letellier et al.
Synthetic Communications, 18, 1975-1975 (1988)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service