Skip to Content
Merck
All Photos(3)

Key Documents

925535

Sigma-Aldrich

Infrared PbS quantum dots

λmax, 1550 nm, 100 mg/mL in toluene

Synonym(s):

Fluorescent nanocrystals, INFIQ® quantum dots, Lead based core type, QDs

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PbS
CAS Number:
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.23

form

liquid

Quality Level

packaging

pkg of 1 mL

concentration

100 mg/mL in toluene

particle size

<10 nm (Nanoparticle)

λmax

≤120 nm (FWHM)
1550 nm

storage temp.

2-8°C

InChI

1S/Pb.S

InChI key

XCAUINMIESBTBL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Lead sulfide quantum dots are inorganic semiconductors with an outer surface coating of organic ligands. Due to their excellent photosensitivity and band-gap tunability, they find applications in photodetectors. These particles are tuned to 1550 nm and are compatible with standard silicon-based complementary metal-oxide semiconductor (CMOS) read-out circuits.

Other applications of these materials include light detection and ranging (LIDAR), 3D laser scanning, solar cells, as well as Time of Flight (ToF) sensing.

Legal Information

Product of Quantum Science Ltd
INFIQ is a registered trademark of Quantum Science Ltd.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 1A - Skin Irrit. 2 - STOT RE 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.2 °F

Flash Point(C)

4 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Integration of PbS Quantum Dot Photodiodes on Silicon for NIR Imaging.
IEEE Sensors Journal, 20(13), 6841-6848 (2020)
G L Whitworth et al.
Nature photonics, 15(10), 738-742 (2021-10-08)
Solution processed semiconductor lasers have achieved much success across the nanomaterial research community, including in organic semiconductors1,2, perovskites3,4 and colloidal semiconductor nanocrystals5,6. The ease of integration with other photonic components, and the potential for upscaling using emerging large area fabrication
Maxim S Gaponenko et al.
Applied optics, 45(3), 536-539 (2006-02-09)
PbS quantum-dot-doped glasses are demonstrated as saturable absorber Q-switches for 2 microm holmium lasers. Q-switched pulses from Ho3+:Y3Sc2Al3O12 and Ho3+:Y3Al5O12 lasers of 50 and 70 ns in duration, respectively, have been obtained demonstrating up to 13% of energy conversion efficiency

Articles

Infrared lead sulfide (PbS)-based quantum dots absorb and emit light across the near-infrared and short-wave infrared wavelengths. Our PbS quantum dots have well-characterized optoelectronic properties and surface chemistry suitable for integration into photodetectors, photovoltaics, and infrared LEDs.

Infrared lead sulfide (PbS)-based quantum dots absorb and emit light across the near-infrared and short-wave infrared wavelengths. Our PbS quantum dots have well-characterized optoelectronic properties and surface chemistry suitable for integration into photodetectors, photovoltaics, and infrared LEDs.

Infrared lead sulfide (PbS)-based quantum dots absorb and emit light across the near-infrared and short-wave infrared wavelengths. Our PbS quantum dots have well-characterized optoelectronic properties and surface chemistry suitable for integration into photodetectors, photovoltaics, and infrared LEDs.

Infrared lead sulfide (PbS)-based quantum dots absorb and emit light across the near-infrared and short-wave infrared wavelengths. Our PbS quantum dots have well-characterized optoelectronic properties and surface chemistry suitable for integration into photodetectors, photovoltaics, and infrared LEDs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service