924288
Copper indium sulfide zinc sulfide (CuInS2/ZnS) core/shell Quantum Dots
in toluene, fluorescence λem 560 nm, 1 mg/mL
Synonym(s):
CIS QD, Fluorescent Nanocrystals, QDs
About This Item
Recommended Products
grade
for analytical purposes
Quality Level
concentration
in toluene
1 mg/mL
fluorescence
FWHM <125 nm
λem 560 nm
, quantum yield ≥40%
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
CuInS2/ZnS quantum dots possess brilliant quantum yield and excellent long-term stability, and these non-toxic quantum dots lack the environmental hazards of cadmium-based quantum dots.
Application
- Batteries
- Photovoltaics
- Light-emitting diodes
- Fluorescent labeling
- Sensing
- Biological imaging systems
Caution
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
39.9 °F
Flash Point(C)
4.4 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Colloidal quantum dots are nano-meter sized, single-crystalline semiconductor fragments that can have highly customizable optical, electronic, and chemical properties through controlled synthesis.
Colloidal quantum dots are nano-meter sized, single-crystalline semiconductor fragments that can have highly customizable optical, electronic, and chemical properties through controlled synthesis.
Colloidal quantum dots are nano-meter sized, single-crystalline semiconductor fragments that can have highly customizable optical, electronic, and chemical properties through controlled synthesis.
Colloidal quantum dots are nano-meter sized, single-crystalline semiconductor fragments that can have highly customizable optical, electronic, and chemical properties through controlled synthesis.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service