909343
2-(4-Fluorophenyl)-5-methylpyridine
≥95%
Synonym(s):
p-F(Me)ppy
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C12H10FN
CAS Number:
Molecular Weight:
187.21
MDL number:
UNSPSC Code:
12352101
Assay
≥95%
form
powder or crystals
reaction suitability
reaction type: Photocatalysis
reagent type: catalyst
mp
58-59 °C
InChI
1S/C12H10FN/c1-9-2-7-12(14-8-9)10-3-5-11(13)6-4-10/h2-8H,1H3
InChI key
WSVYYUKIXFSDTE-UHFFFAOYSA-N
Application
2-(4-Fluorophenyl)-5-methylpyridine is a ligand used for the preparation of Ir(III) photocatalysts.
Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)
Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Jonathan A Porras et al.
Journal of the American Chemical Society, 138(30), 9460-9472 (2016-07-09)
A series of fluorinated Ir(III)-terpyridine-phenylpyridine-X (X = anionic monodentate ligand) complexes were synthesized by selective C-F activation, whereby perfluorinated phenylpyridines were readily complexed. The combination of fluorinated phenylpyridine ligands with an electron-rich tri-tert-butyl terpyridine ligand generates a "push-pull" force on
Yufan Liang et al.
Nature, 559(7712), 83-88 (2018-06-22)
Over the past three decades, considerable progress has been made in the development of methods to construct sp2 carbon-nitrogen (C-N) bonds using palladium, copper or nickel catalysis1,2. However, the incorporation of alkyl substrates to form sp3 C-N bonds remains one
Andrew G Capacci et al.
Nature chemistry, 9(11), 1073-1077 (2017-10-25)
Although the α-alkylation of ketones has already been established, the analogous reaction using aldehyde substrates has proven surprisingly elusive. Despite the structural similarities between the two classes of compounds, the sensitivity and unique reactivity of the aldehyde functionality has typically
Yong Yao Loh et al.
Science (New York, N.Y.), 358(6367), 1182-1187 (2017-11-11)
Deuterium- and tritium-labeled pharmaceutical compounds are pivotal diagnostic tools in drug discovery research, providing vital information about the biological fate of drugs and drug metabolites. Herein we demonstrate that a photoredox-mediated hydrogen atom transfer protocol can efficiently and selectively install
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service