Skip to Content
Merck
All Photos(3)

Documents

90660

Sigma-Aldrich

Tribenzylamine

≥99.0% (NT)

Synonym(s):

TBA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5CH2)3N
CAS Number:
Molecular Weight:
287.40
Beilstein:
2214682
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (NT)

form

powder

mp

91-94 °C (lit.)

SMILES string

C(N(Cc1ccccc1)Cc2ccccc2)c3ccccc3

InChI

1S/C21H21N/c1-4-10-19(11-5-1)16-22(17-20-12-6-2-7-13-20)18-21-14-8-3-9-15-21/h1-15H,16-18H2

InChI key

MXHTZQSKTCCMFG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tribenzylamine (TBA) is a tertiary amine which can be used :
  • As a nitrogen group source for the reactions involving C−N bond formation.
  • For the synthesis of imine i.e. N−benzylidene benzylamine by aerobic oxidative condensation.
  • As an extractant for the separation and determination of Cr(VI) and Cr(III) from wastewater.

TBA can also undergo debenzylation in the presence of ceric ammonium nitrate (CAN) to form dibenzylamine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

399.2 °F - closed cup

Flash Point(C)

204 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly active and selective gold catalysts for the aerobic oxidative condensation of benzylamines to imines and one-pot, two-step synthesis of secondary benzylamines.
Grirrane A, et al.
J. Catal., 264(2), 138-144 (2009)
Chemoselective oxidative debenzylation of tertiary N-benzyl amines.
Bull SD, et al.
Chemical Communications (Cambridge, England), 165(5), 337-338 (2000)
Copper-Catalyzed Oxidative Amination of Benzoxazoles via C- H and C- N Bond Activation: A New Strategy for Using Tertiary Amines as Nitrogen Group Sources.
Guo S, et al.
Organic Letters, 13(3), 522-525 (2010)
Céline Burnier et al.
Talanta, 192, 135-141 (2018-10-24)
Nowadays, Gas Chromatography Mass Spectrometry (GC-MS) is mainly used in forensic sciences but suffers from limitations when the analysed compounds are thermally instable as it is the case for THC-A (Tetrahydrocannabinolic Acid) which is converted into Δ9-THC (Δ9-Tetrahydrocannabinol) that subsequently
Yan Qu et al.
Zeitschrift fur Naturforschung. C, Journal of biosciences, 58(9-10), 640-642 (2003-10-28)
Two compounds, (p-methoxyphenyl) diphenylmethanol (1) and tribenzylamine (2), were isolated from Humulus lupulus. Their structures were established on the basis of spectral evidence (MS, IR, NMR, HMBC, HMQC, 1H-1H COSY experiments). Compounds 1 and 2 were found as natural products

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service