Skip to Content
Merck
All Photos(3)

Key Documents

904848

Sigma-Aldrich

2-Chloro-1,3-bis(2,6-diisopropylphenyl)imidazolium chloride

≥95%

Synonym(s):

1,3-Bis(2,6-di-i-propylphenyl)-2-chloroimidazolium chloride, Chloroimidazolium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H36Cl2N2
CAS Number:
Molecular Weight:
459.49
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

Assay

≥95%

form

powder

functional group

chloro

SMILES string

[Cl+]=C1N(C=CN1c3c(cccc3C(C)C)C(C)C)c2c(cccc2C(C)C)C(C)C.[Cl-]

InChI

1S/C27H36ClN2.ClH/c1-17(2)21-11-9-12-22(18(3)4)25(21)29-15-16-30(27(29)28)26-23(19(5)6)13-10-14-24(26)20(7)8;/h9-20H,1-8H3;1H/q+1;/p-1

InChI key

JDMACANGISWEGX-UHFFFAOYSA-M

Application

This chloroimidazolium chloride is used in the deoxyfluorination of phenols. Recently, the Ritter lab used it in conjunction with 902314 and 906794 to incorporate the [18F]fluoride radiolabel site specifically into polypeptides for use with positron emission tomography (PET) imaging in the study of various biological applications.

Automate your fluorination reactions with Synple Automated Synthesis Platform (SYNPLE-SC002)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Martin G Strebl et al.
ACS central science, 3(9), 1006-1014 (2017-10-06)
Histone deacetylase 6 (HDAC6) function and dysregulation have been implicated in the etiology of certain cancers and more recently in central nervous system (CNS) disorders including Rett syndrome, Alzheimer's and Parkinson's diseases, and major depressive disorder. HDAC6-selective inhibitors have therapeutic
Robert J Gilliard et al.
Chemical communications (Cambridge, England), 53(91), 12325-12328 (2017-11-03)
The reaction of the chloroimidazolium chloride salt, [NHC-Cl][Cl], NHC = C{N(2,6-iPr2C6H3)CH}2 (1) with two equivalents of sodium phosphaethynolate, Na[OCP]·(dioxane)2.5, results in the formation of NHC-{cyclo-(CO)-P2-C(O)} (2) and NHC-P2-C(O)-NHC (3). Notably, in the presence of free NHC ligand, compound 2 converts
Daniel Mendoza-Espinosa et al.
Journal of the American Chemical Society, 132(21), 7264-7265 (2010-05-07)
Using the nucleophilicity of NHCs and aNHCs, as well as the leaving group ability of the former, the carbon-carbon double bond of imidazol-2-ylidenes can be readily mono- and difunctionalized. These results provide also a new light on the formation of
M Hassan Beyzavi et al.
ACS central science, 3(9), 944-948 (2017-10-06)
The deficiency of robust and practical methods for 18F-radiofluorination is a bottleneck for positron emission tomography (PET) tracer development. Here, we report the first transition-metal-assisted 18F-deoxyfluorination of phenols. The transformation benefits from readily available phenols as starting materials, tolerance of
Teppei Fujimoto et al.
Organic process research & development, 18(8), 1041-1044 (2014-09-02)
We report a practical synthesis method of the reagent PhenoFluor on decagram scale, provide a new formulation of PhenoFluor as a toluene solution, which should decrease challenges associated with the moisture sensitivity of the reagent, and expand the substrate scope

Related Content

The Ritter lab currently focuses on fluorination chemistry for late-stage functionalization of complex natural and unnatural products. PhenoFluor™ has been developed as a general reagent for the selective, predictable, direct deoxyfluorination of complex alcohols and phenols.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service