Skip to Content
Merck
All Photos(3)

Documents

901434

Sigma-Aldrich

Methylammonium iodide

greener alternative

≥99%, anhydrous

Synonym(s):

Methanamine, hydriodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH6IN
CAS Number:
Molecular Weight:
158.97
UNSPSC Code:
12352302
NACRES:
NA.23

grade

anhydrous

Quality Level

Assay

≥99%

form

crystals

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

InChI

1S/CH5N.HI/c1-2;/h2H2,1H3;1H

InChI key

LLWRXQXPJMPHLR-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.

Application

Methylammonium iodide (MAI) has been widely used in the development of perovskite solar cells. MAI is typically used as a precursor material in the fabrication of perovskite thin films. It acts as a source of methylammonium cations (CH3NH3+) and iodide anions (I-) that are necessary for the formation of the perovskite crystal structure.
Methylammonium iodide, an organic halide based perovskite material, can be used in the fabrication of high performance organic solar cells.
Organohalide based perovskites have emerged as an important class of material for solar cell applications. Our perovskites precursors with extremely low water contents are useful for synthesizing mixed cation or anion perovskites needed for the optimization of the band gap, carrier diffusion length and power conversion efficiency of perovskites based solar cells.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deciphering the role of impurities in methylammonium iodide and their impact on the performance of perovskite solar cells
Levchuk I, et al.
Advanced Materials Interfaces, 3(22), 1600593-1600593 (2016)
Spectroscopic study on the impact of methylammonium iodide loading time on the electronic properties in perovskite thin films
Cheng Y, et al.
Journal of Material Chemistry A, 4(2), 561-567 (2016)
Recent Advances in Hybrid Halide Perovskites-based Solar Cells.
Kalyanasundaram K, et al.
Material Matters , 11, 3-3 (2016)
Crystallization of a perovskite film for higher performance solar cells by controlling water concentration in methyl ammonium iodide precursor solution
Adhikari N, et al.
Nanoscale, 8(5), 2693-2703 (2016)
Entropic stabilization of mixed A-cation ABX3 metal halide perovskites for high performance perovskite solar cells.
Yi C, et al.
Energy & Environmental Science, 9, 656-656 (2016)

Articles

To achieve net-zero emissions by 2050, renewable power contributions must triple. Photovoltaic stations provide vital utility power, achieved primarily through third- and fourth-generation technology. Promising trends include recycling and revolutionary, ultra-lightweight, flexible, and printable solar cells.

To achieve net-zero emissions by 2050, renewable power contributions must triple. Photovoltaic stations provide vital utility power, achieved primarily through third- and fourth-generation technology. Promising trends include recycling and revolutionary, ultra-lightweight, flexible, and printable solar cells.

To achieve net-zero emissions by 2050, renewable power contributions must triple. Photovoltaic stations provide vital utility power, achieved primarily through third- and fourth-generation technology. Promising trends include recycling and revolutionary, ultra-lightweight, flexible, and printable solar cells.

A brief tutorial on alternative energy materials for advanced batteries and fuel cells, as well as high-purity inorganics, conducting polymers, and electrolytes.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service