Skip to Content
Merck
All Photos(2)

Key Documents

901112

Sigma-Aldrich

PhenN O-PC B0301

New Iridium, ≥97%

Synonym(s):

Miyake polymerization organophotoredox catalyst, 5,10-Di(2-Naphthyl)-5,10-dihydrophenazine, PhenN_2Naph

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H22N2
CAS Number:
Molecular Weight:
434.53
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

Quality Level

Assay

≥97%

form

powder or crystals

reaction suitability

reagent type: catalyst
reaction type: Photocatalysis

photocatalyst activation

460 nm

SMILES string

[n]3(c4c([n](c7c3cccc7)c5cc6c(cc5)cccc6)cccc4)c1cc2c(cc1)cccc2

InChI key

HPIOBCHPZVAATK-UHFFFAOYSA-N

Application

This dihydrophenazine-based organic photoredox catalyst (in addition to the dihydrophenazine catalyst 901111) was designed to be a strong excited-state reductant and possesses advanced photophysical and electrochemical properties, enabling it to serve as a sustainable replacement for ruthenium- or iridium-based photoredox catalysts. For example, dihydrophenazine and phenoxazine derivatives were demonstrated to replace ruthenium or iridium complexes in the application of photoredox-catalyzed atom transfer radical polymerization (ATRP) for controlled polymer synthesis and small molecule transformations such as trifluoromethylation, atom transfer radical addition, and dual Nickel/photoredox catalyzed C-N and C-S cross-couplings. Dihydrophenazine- and phenoxazine-based organic photoredox catalysts were introduced in collaboration with the Miyake Research Group.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Legal Information

Patent application PCT/US2016/058245. Sold in collaboration with New Iridium Inc. For orders greater than 25g, please contact New Iridium at [email protected] or visit https://www.newiridium.com.
PhenN O-PC is a trademark of New Iridium Inc.
Phenox O-PC is a trademark of New Iridium LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryan M Pearson et al.
Journal of the American Chemical Society, 138(35), 11399-11407 (2016-08-25)
N-Aryl phenoxazines have been synthesized and introduced as strongly reducing metal-free photoredox catalysts in organocatalyzed atom transfer radical polymerization for the synthesis of well-defined polymers. Experiments confirmed quantum chemical predictions that, like their dihydrophenazine analogs, the photoexcited states of phenoxazine
Jordan C Theriot et al.
Science (New York, N.Y.), 352(6289), 1082-1086 (2016-04-02)
Atom transfer radical polymerization (ATRP) has become one of the most implemented methods for polymer synthesis, owing to impressive control over polymer composition and associated properties. However, contamination of the polymer by the metal catalyst remains a major limitation. Organic
Ya Du et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 23(46), 10962-10968 (2017-06-28)
Photoredox catalysis is a versatile approach for the construction of challenging covalent bonds under mild reaction conditions, commonly using photoredox catalysts (PCs) derived from precious metals. As such, there is need to develop organic analogues as sustainable replacements. Although several
Chern-Hooi Lim et al.
Journal of the American Chemical Society, 139(1), 348-355 (2016-12-16)
Photoexcited intramolecular charge transfer (CT) states in N,N-diaryl dihydrophenazine photoredox catalysts are accessed through catalyst design and investigated through combined experimental studies and density functional theory (DFT) calculations. These CT states are reminiscent of the metal to ligand charge transfer

Articles

Photoredox catalysis is a powerful synthetic methodology to form challenging covalent bonds using light irradiation. It is effective for light-driven polymer and small molecule synthesis.

Related Content

Research in the Miyake laboratory focuses on catalysis, polymer chemistry, and materials science.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service