Skip to Content
Merck
All Photos(2)

Key Documents

900656

Sigma-Aldrich

Methoxy poly(ethylene glycol)-b-poly(L-lactide)

5k-10k

Synonym(s):

mPEG-b-PLA, mPEG-PLA, mPEG-PLLA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO[CH(CH3)COO]m[CH2CH2O]nCH3
UNSPSC Code:
12162002
NACRES:
NA.23

Quality Level

form

solid

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Biocompatible, amphiphilic block copolymer composed of a hydrophilic PEG block and a hydrophobic poly(L-lactide) (PLLA) block. These materials have been used in control release and nanoparticle formulation for drug encapsulation and delivery applications. Well-defined materials with varying properties can be prepared by controlling the relative length of each polymer block. Hydroxyl termination allows for facile further chemical modification of these materials.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.7 °F

Flash Point(C)

> 110.4 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Injectable biodegradable polymeric system for preserving the active form and delayed-release of camptothecin anticancer drugs.
Mert O, et al.
Royal Society of Chemistry Advances, 2(1), 176-185 (2012)
Zhigang Xie et al.
Journal of biomedical materials research. Part A, 88(1), 238-245 (2008-02-21)
Docetaxel (DX) is one of the most effective antineoplastic drugs. Its current clinical administration is limited because of its hydrophobicity and serious side effects. A polymer/DX conjugate is designed and successfully prepared to solve these problems. It is monomethoxy-poly(ethylene glycol)-block-poly(L-lactide)/DX

Articles

The development of drugs that target specific locations within the human body remains one of the greatest challenges in biomedicine today.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service