Skip to Content
Merck
All Photos(1)

Key Documents

85730

Sigma-Aldrich

Stearoyl chloride

technical, ≥90% (GC), strongly brown

Synonym(s):

Octadecanoyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)16COCl
CAS Number:
Molecular Weight:
302.92
Beilstein:
639784
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Assay

≥90% (GC)

form

liquid

color

strongly brown

refractive index

n20/D 1.454 (lit.)

bp

174-178 °C/2 mmHg (lit.)

mp

21-22 °C (lit.)

density

0.897 g/mL at 25 °C (lit.)

functional group

acyl chloride

SMILES string

CCCCCCCCCCCCCCCCCC(Cl)=O

InChI

1S/C18H35ClO/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h2-17H2,1H3

InChI key

WTBAHSZERDXKKZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Stearoyl chloride can modify cellulose diacetate (CDA) by replacing residual free hydroxyl groups in synthesizing materials that are used to produce surgical masks, smocks and other industrial products to absorb dust and haze-fog. It can be used in the synthesis of ultrastable coaxial cable-like superhydrophobic mesh. It is also used in the synthesis of stearoyl modified chitosan as drug vehicles for paclitaxel delivery.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

329.0 °F - closed cup

Flash Point(C)

165 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amphiphilic nanoparticles based on poly (vinyl pyrrolidone) and stearoyl modified chitosan as drug vehicles for paclitaxel delivery.
Liu Z, et al.
Materials Letters, 185, 226-229 (2016)
Ultrastable coaxial cable-like superhydrophobic mesh with self-adaption effect: facile synthesis and oil/water separation application.
Xiao C, et al.
Journal of Material Chemistry A, 4(21), 8080-8090 (2016)
Çağla Koşak Söz et al.
ACS applied materials & interfaces, 10(43), 37478-37488 (2018-10-27)
We introduce the design of Janus-type paper sheets where one side of the paper exhibits superhydrophobic properties, whereas the other side of the sheet remains hydrophilic and therefore can take up aqueous solutions by capillary wicking. Such papers are being
Acylation of Cellulose Diacetate by Stearoyl Chloride and Characterization for the Acylated Product.
Wang Q, et al.
J. Polym. Mater., 32(4) (2015)
Shuangfei Wang et al.
Carbohydrate polymers, 195, 39-44 (2018-05-29)
Superhydrophobic surfaces have attracted great attention due to their attractive properties. Biopolymer-based low-cost and environmentally-friendly superhydrophobic coatings with easy-to-perform fabrication methods are always desirable. Herein, we report superhydrophobic surfaces using a one-step spray-coating of chitosan-based nanoparticles. The particles were easily

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service