Skip to Content
Merck
All Photos(1)

Key Documents

83000

Sigma-Aldrich

Potassium methanesulfonate

≥98.0% (dry substance, T)

Synonym(s):

Methanesulfonic acid potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH3KO3S
CAS Number:
Molecular Weight:
134.20
Beilstein:
3914899
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (dry substance, T)

form

crystals

loss

<10% loss on drying

functional group

sulfonic acid

SMILES string

[K+].CS([O-])(=O)=O

InChI

1S/CH4O3S.K/c1-5(2,3)4;/h1H3,(H,2,3,4);/q;+1/p-1

InChI key

XWIJIXWOZCRYEL-UHFFFAOYSA-M

General description

1,3,(6,7)-Naphthalenetrisulfonic acid trisodium salt hydrate is an anionic chromophore often used in the fabrication of Ni, PPy-NTS and PPy-PTS electrodes to enhance the hydrogen evolution reaction.

Application

Potassium methanesulfonate can be employed as a cocatalyst in ruthenium-catalyzed C-H arylation of heteroarylbenzenes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Studies on the Cocatalyst in Ruthenium-Catalyzed C-H Arylation
Seki M, et al.
Synthesis, 47(10), 1423-1435 (2015)
Sara Ibañez et al.
Frontiers in computational neuroscience, 13, 89-89 (2020-02-06)
Behavioral studies have shown spatial working memory impairment with aging in several animal species, including humans. Persistent activity of layer 3 pyramidal dorsolateral prefrontal cortex (dlPFC) neurons during delay periods of working memory tasks is important for encoding memory of
Chandra Prajapati et al.
Biology open, 7(7) (2018-07-05)
Human induced pluripotent stem cell derived cardiomyocytes (hiPSC-CMs) offer a unique in vitro platform to study cardiac diseases, as they recapitulate many disease phenotypes. The membrane potential (Vm) and intracellular calcium (Ca2+) transient (CaT) are usually investigated separately, because incorporating
Ning Song et al.
Implant dentistry, 28(1), 54-61 (2018-12-07)
Titanium (Ti) is the key material used in dental implants because of its excellent biocompatibility. But wear and corrosion Ti particles had been widely reported to induce inflammation and promote bone absorption. However, little information is known about the damage
Benoit Labonté et al.
Nature medicine, 23(9), 1102-1111 (2017-08-22)
Major depressive disorder (MDD) is a leading cause of disease burden worldwide. While the incidence, symptoms and treatment of MDD all point toward major sex differences, the molecular mechanisms underlying this sexual dimorphism remain largely unknown. Here, combining differential expression

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service