Skip to Content
Merck
All Photos(3)

Key Documents

795755

Sigma-Aldrich

n-Dodecylphosphonic acid

Synonym(s):

DDPA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H27O3P
CAS Number:
Molecular Weight:
250.31
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

solid

Quality Level

mp

68-73 °C

SMILES string

CCCCCCCCCCCCP(O)(O)=O

InChI

1S/C12H27O3P/c1-2-3-4-5-6-7-8-9-10-11-12-16(13,14)15/h2-12H2,1H3,(H2,13,14,15)

InChI key

SVMUEEINWGBIPD-UHFFFAOYSA-N

General description

Styryl 9M is an organic laser dye that is used as a non-linear optical(NLO) component. It is biocompatible in nature and can be used as a chromophore for photon absorption based microscopy.

Application

Styryl 9M may be used as a near-infrared based dyes that can be incorporated with amino functionalized polystyrene nanoparticles for the tracing and tracking of tumor cells. It exhibits non-linear optical properties which may be used in the fabrication of photonic crystal for application as ultrafast optical switches.
Tuning of surface energy, wetting, and work function of metal oxides

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Environment-sensitive two-photon dye
Linear and Nonlinear Optics of Organic Materials VIII, 7049, 70490Y-70490Y (2008)
Long-chain functional bisphosphonates: synthesis, anticalcification, and antiresorption activity
Meziane, D.;
Heteroatom Chem., 20, 369-377 (2009)
Ultra-fast optical switches using 1D polymeric photonic crystals.
Katouf R, et al.
Ultra-fast optical switches using 1D polymeric photonic crystals, 3(2-3), 116-119 (2005)
Target-specific nanoparticles containing a broad band emissive NIR dye for the sensitive detection and characterization of tumor development.
Behnke T, et al.
Biomaterials, 34(1), 160-170 (2013)
Fredric G Svensson et al.
Materials (Basel, Switzerland), 13(21) (2020-11-04)
Titania (anatase) nanoparticles were anisotropically functionalized in water-toluene Pickering emulsions to self-assemble into nanoshells with diameters from 500 nm to 3 μm as candidates for encapsulation of drugs and other compounds. The water-phase contained a hydrophilic ligand, glucose-6-phosphate, while the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service