Skip to Content
Merck
All Photos(1)

Documents

793353

Sigma-Aldrich

Tungsten oxide (WO3-x) nanoparticle ink

Synonym(s):

Avantama P-10, Nanograde P-10, Tungsten oxide nanoparticle dispersion, Tungsten oxide suspension, WO3 dispersion, WO3 ink

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
WO3-x
UNSPSC Code:
12352103
NACRES:
NA.23

form

dispersion

Quality Level

concentration

2.5 wt. % in 2-propanol

particle size

<50 nm (BET)

density

0.7992 g/mL at 25 °C

General description

This WO3-x nanoparticle ink is for slot-dye, spin-coating and doctor blading for the use as hole transport layer in printed electronics. Tungsten oxide nanoparticle ink is a hole-selective interface layer ink based on a colloidal suspension of tungsten oxide (WO3) nanoparticles in isopropanol. The average size of WO3 particle is optimized around 12-16 nm. Tungsten oxide nanoparticle exhibits high work function, processability and easy layer formation on hydrophilic as well as hydrophobic substrates.This WO3-x nanoparticle ink is universally applicable in normal and inverted architecture solar cells.
Annealing temperature <100°C.

Application

WO3 nanoparticle ink can be applied in OPV cells as hole extraction layer (HEL) materials. Tungsten oxide nanoparticle ink can be mixed with PEDOT:PSS formulations in order to fine tune electronic and morphological dry layer properties (e.g. conductivity, surface roughness or layer porosity).

Other Notes

Prior to application: Ultrasonicate and (optionally) filter through 0.45 μm PTFE filter
Working conditions: Application and film drying under nitrogen (or low humidity)
Post-treatment: Annealing of deposited WO3-x films at 80°C - 120°C

Legal Information

Product of Avantama Ltd.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inverted structure organic photovoltaic devices employing a low temperature solution processed WO3 anode buffer layer
Christoph J. Brabec; et al.
Organic Electronics, 13(11), 2479-2484 (2012)
High Fill Factor Polymer Solar Cells Incorporating a Low Temperature Solution Processed WO3 Hole Extraction Layer
Christoph J. Brabec; et al.
Advanced Energy Materials, 2, 1433-1438 (2012)
Chun-Chao Chen et al.
Advanced materials (Deerfield Beach, Fla.), 26(32), 5670-5677 (2014-07-22)
Tandem solar cells have the potential to improve photon conversion efficiencies (PCEs) beyond the limits of single-junction devices. In this study, a triple-junction tandem design is demonstrated by employing three distinct organic donor materials having bandgap energies ranging from 1.4
Flexible organic tandem solar modules with 6% efficiency: combining roll-to-roll compatible processing with high geometric fill factors
Energy & Environmental Science, 7, 3284?3290-3284?3290 (2014)
Lin Zhou et al.
Scientific reports, 9(1), 8778-8778 (2019-06-21)
This paper presents perovskite solar cells employed with WO3 nanoparticles embedded carbon top electrode. WO3 nanoparticles works as an inorganic hole-transport material (HTM) to promote the hole-extraction in the perovskite/carbon interface as revealed by efficiency, electrochemical impedance and external quantum

Articles

Find advantages of inorganic interface layer inks for organic electronic & other applications.

Professors Tokito and Takeda share design principles and optimization protocols for organic electronic devices, focusing on flexibility and low cost.

Professors Tokito and Takeda share design principles and optimization protocols for organic electronic devices, focusing on flexibility and low cost.

Professors Tokito and Takeda share design principles and optimization protocols for organic electronic devices, focusing on flexibility and low cost.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service