Skip to Content
Merck
All Photos(1)

Key Documents

791806

Sigma-Aldrich

Yu-Wasa Auxiliary

97%

Synonym(s):

2,3,5,6-Tetrafluoro-4-(trifluoromethyl)aniline, 2,3,5,6,α,α,α-Heptafluoro-p-toluidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C6F4NH2
CAS Number:
Molecular Weight:
233.09
Beilstein:
2657893
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

reaction suitability

reaction type: C-C Bond Formation
reagent type: catalyst
reaction type: C-H Activation

refractive index

n20/D 1.431 (lit.)
n20/D 1.432

bp

186 °C (lit.)

density

1.662 g/mL at 25 °C
1.687 g/mL at 25 °C (lit.)

SMILES string

Nc1c(F)c(F)c(c(F)c1F)C(F)(F)F

InChI

1S/C7H2F7N/c8-2-1(7(12,13)14)3(9)5(11)6(15)4(2)10/h15H2

InChI key

FJOACTZFMHZHSC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Acidic amides are superior directing groups for promoting C-H activation reactions with both Pd(0)/PR3 and Pd(II) catalysts.

Used in the Preparation of
  • Lactams via palladium-catalyzed olefination of arylamides with benzylacrylate, followed by 1,4-conjugate addition
  • N-(fluorinated aryl)benzamides as substrates for regioselective C-H amination reactions with O-benzoylhydroxylamines
  • Substituted succinimides via palladium-catalyzed carbonylation of N-aryl amides
  • N-aryl cyclopropanecarboxamide substrates and various amino acid ligands for palladium-catalyzed C-H activation of cyclopropanes

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service