Skip to Content
Merck
All Photos(1)

Key Documents

790346

Sigma-Aldrich

ITO

30 nm (SEM)

Synonym(s):

Indium tin oxide, ITO

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

nanopowder

Quality Level

composition

In2O3, 90%
SnO2, 10%

particle size

30 nm (SEM)

mp

1910 °C (lit.)

density

1.2 g/mL at 25 °C (lit.)

SMILES string

O=[SnH2].O=[In]O[In]=O.O=[In]O[In]=O.O=[In]O[In]=O.O=[In]O[In]=O.O=[In]O[In]=O.O=[In]O[In]=O.O=[In]O[In]=O.O=[In]O[In]=O.O=[In]O[In]=O

InChI

1S/2In.5O.Sn

InChI key

LNNWKAUHKIHCKO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Indium Tin oxide nanoparticles have been investigated for use in the following:
  • transparent electrodes(1)
  • the formation of conductive paper and organic contaminants (2)
  • the formation of transparent conductive oxide films and magnetic nanocomposites(3)
  • electrochromic materials and as well as in biomedical applications(4)

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Conductive paper fabricated by layer-by-layer assembly of polyelectrolytes and ITO nanoparticles.
Peng CQ, et al.
Nanotechnology, 19(50) (2008)
Characterization of dip-coated ITO films derived from nanoparticles synthesized by?low-pressure spray pyrolysis.
Ogi T, et al.
Journal of Nanoparticle Research, 8(3-4), 343-350 (2006)
Progress in Electrochromics and Thermochromics: Two New Applications Involving ITO Nanoparticles.
Li S, et al.
Society of Vacuum Coaters; 55th Annual Technical Conference Proceedings, 41-46 (2012)
Generation of transparent conductive electrodes by laser consolidation of LIFT printed ITO nanoparticle layers.
Baum M, et al.
Applied Physics. A, Materials Science & Processing, 111(3), 799-805 (2013)

Articles

TiO2 exhibits wide band gap semiconductor and memristor properties electronically, with high opacity and UV absorbance optically.

TiO2 exhibits wide band gap semiconductor and memristor properties electronically, with high opacity and UV absorbance optically.

TiO2 exhibits wide band gap semiconductor and memristor properties electronically, with high opacity and UV absorbance optically.

TiO2 exhibits wide band gap semiconductor and memristor properties electronically, with high opacity and UV absorbance optically.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service