Skip to Content
Merck
All Photos(3)

Key Documents

778095

Sigma-Aldrich

Silver nanowires

diam. × L 200  nm × 10 μm, 0.5% (isopropyl alcohol suspension)

Synonym(s):

Silver nanofibers, Silver nanowhiskers, Silver nanowire

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ag
CAS Number:
Molecular Weight:
107.87
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

form

liquid (suspension)
nanowires

Quality Level

concentration

0.5% (isopropyl alcohol suspension)

diam. × L

200  nm × 10 μm

density

0.779 g/mL at 25 °C

SMILES string

[Ag]

InChI

1S/Ag

InChI key

BQCADISMDOOEFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Silver nanowires are a one-dimensional nanomaterial of metallic silver with a high aspect ratio (length vs diameter). These nanowires have a length of 10 microns and a diameter of 200 nm. Our silver nanowires combine high electrical conductivity with low light extinction in the visible making them useful in a wide range of applications, from transparent electrodes, to temperature and pressure sensors.

Application

Silver nanowires have been studied for use in electronic and biological applications including transparent conductors.[1]; and water sterilization.[2] . Silver nanowires have also been studied as plasmonic[3] and as components in amorphous semiconductors.[4].

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sukanta De et al.
ACS nano, 3(7), 1767-1774 (2009-06-26)
We have used aqueous dispersions of silver nanowires to prepare thin, flexible, transparent, conducting films. The nanowires are of length and diameter close to 6.5 μm and 85 nm, respectively. At low thickness, the films consist of networks but appear
Luo; J.;
Advanced Materials, 16(17), 1512-1515 (2004)
David T Schoen et al.
Nano letters, 10(9), 3628-3632 (2010-08-24)
The removal of bacteria and other organisms from water is an extremely important process, not only for drinking and sanitation but also industrially as biofouling is a commonplace and serious problem. We here present a textile based multiscale device for
Aric W Sanders et al.
Nano letters, 6(8), 1822-1826 (2006-08-10)
We report the coupling of free-space photons (vacuum wavelength of 830 nm) to surface plasmon modes of a silver nanowire. The launch of propagating plasmons, and the subsequent emission of photons, is selective and occurs only at ends and other
Ana López-Serrano Oliver et al.
Environmental pollution (Barking, Essex : 1987), 189, 87-91 (2014-03-20)
Silver nanoparticles (AgNPs) are widely used in many applications and likely released into the aquatic environment. There is increasing evidence that Ag is efficiently delivered to aquatic organisms from AgNPs after aqueous and dietary exposures. Accumulation of AgNPs through the

Articles

The ability to pattern conductive electrodes is technologically relevant for several applications, including photovolatics, displays, sensors, and biomedical devices.

Silver nanomaterials have unique physical, chemical, and optical properties that are currently being leveraged for a wide variety of biological applications.

Among various ceramics, one-dimensional (1-D) piezoelectric ceramics have attracted significant scientific attention for use in energy harvesting.

TiO2 exhibits wide band gap semiconductor and memristor properties electronically, with high opacity and UV absorbance optically.

See All

Questions

  1. 778095-25ML 제품 형태가 IPA 에 silver nanowire 가 녹아있는 용액으로 25 ml 에 ₩478,877 인지 궁금합니다.

    1 answer
    1. Yes, this product is a 0.5% silver nanowire solution in Isopropanol. Pricing and currency vary from country to customer. To verify pricing, please navigate to the link https://www.sigmaaldrich.com/support/customer-support, click on "Products Pricing & Availability" under the Products Section with all the required information so that a member of the Customer Service team can reach out to assist further. Thank you.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service