Skip to Content
Merck
All Photos(1)

Key Documents

773875

Sigma-Aldrich

Platinum, nanoparticle dispersion

3 nm particle size, 1,000 ppm in H2O, 99.99% trace metals basis

Synonym(s):

Platinum colloid, Pt NP, Pt dispersion

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Pt
Molecular Weight:
195.08
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.99% trace metals basis

form

dispersion
nanoparticle

reaction suitability

reagent type: catalyst
core: platinum

concentration

1,000 ppm in H2O

particle size

3 nm

density

0.997 g/mL at 25 °C

SMILES string

[Pt]

InChI

1S/Pt

InChI key

BASFCYQUMIYNBI-UHFFFAOYSA-N

Application

Platinum nanoparticles are used in several important catalytic applications including fuel cells and photocatalysts. Platinum nanocrystal arrays have also been studied as nonvolatile memory systems.

WGK

nwg


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yongyan Mu et al.
The journal of physical chemistry. B, 109(47), 22212-22216 (2006-07-21)
We report a novel process to prepare well-dispersed Pt nanoparticles on CNTs. Pt nanoparticles, which were modified by the organic molecule triphenylphosphine, were deposited on multiwalled carbon nanotubes by the organic molecule, which acts as a cross linker. By manipulating
Lee; C.;
Journal of Electronic Materials, 34, 1-11 (2005)
Wang; X.;
Chemistry (Weinheim An Der Bergstrasse, Germany), 11, 2997-3004 (2005)
L Martinaga Pintarić et al.
Polymers, 12(6) (2020-05-30)
In this research, a textile surface was modified by the sol-gel methodology with a new antimicrobial coating containing nanoparticles active against bacteria resistant to antibiotics. The effect of ultrasonic irradiation power (40 to 90 kHz), the concentration of reagents (nanoparticles

Articles

TiO2 exhibits wide band gap semiconductor and memristor properties electronically, with high opacity and UV absorbance optically.

TiO2 exhibits wide band gap semiconductor and memristor properties electronically, with high opacity and UV absorbance optically.

TiO2 exhibits wide band gap semiconductor and memristor properties electronically, with high opacity and UV absorbance optically.

TiO2 exhibits wide band gap semiconductor and memristor properties electronically, with high opacity and UV absorbance optically.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service