Skip to Content
Merck
All Photos(1)

Key Documents

772453

Sigma-Aldrich

Fmoc-Lys(palmitoyl)-OH

95%

Synonym(s):

Palmitoyl lysine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H54N2O5
CAS Number:
Molecular Weight:
606.84
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Quality Level

Assay

95%

form

powder or crystals

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

mp

71-83 °C

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCC(=O)NCCCC[C@@H](NC(=O)OCC1c2ccccc2-c3ccccc13)C(O)=O

InChI

1S/C37H54N2O5/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-26-35(40)38-27-20-19-25-34(36(41)42)39-37(43)44-28-33-31-23-17-15-21-29(31)30-22-16-18-24-32(30)33/h15-18,21-24,33-34H,2-14,19-20,25-28H2,1H3,(H,38,40)(H,39,43)(H,41,42)/t34-/m1/s1

InChI key

DQZCUQJUXKWZEE-UUWRZZSWSA-N

Application

Amino acid-lipid conjugate for the solid phase synthesis of peptide-lipids. Lipidated peptides have been shown in some cases to lead to increased cellular permeability and metabolic stability.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Joanne T Blanchfield et al.
Journal of medicinal chemistry, 46(7), 1266-1272 (2003-03-21)
The alpha-conotoxin MII is a two disulfide bridge containing, 16 amino acid long peptide toxin isolated from the marine snail Conus magus. This toxin has been found to be a highly selective and potent inhibitor of neuronal nicotinic acetylcholine receptors
Grzegorz Bulaj et al.
Journal of medicinal chemistry, 51(24), 8038-8047 (2008-12-05)
Galanin is an endogenous neuropeptide that modulates seizures in the brain. Because this neuropeptide does not penetrate the blood-brain barrier, we designed truncated galanin analogues in which nonessential amino acid residues were replaced by cationic and/or lipoamino acid residues. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service