Skip to Content
Merck
All Photos(2)

Key Documents

756490

Sigma-Aldrich

Yttrium oxide, europium doped

greener alternative

99% trace metals basis

Synonym(s):

Europium doped yttria, Europium yttrium oxide, Red phosphor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Y1.92Eu0.08O3
CAS Number:
Molecular Weight:
230.85
EC Number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Quality Level

Assay

99% trace metals basis

form

powder

composition

Y1.92Eu0.08O3

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

particle size

4-8 μm

mp

>1600 °C (lit.)

fluorescence

λex 611 nm; λem ±2 nm (UV excitation source)

greener alternative category

SMILES string

[Eu].O=[Y]O[Y]=O

InChI

1S/Eu.3O.2Y

InChI key

CVLPANXSBBGVHI-UHFFFAOYSA-N

General description

Color coordinate, x: 0.645 +/- 0.015
Color coordinate, y: 0.345 +/- 0.015
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.
Yttrium oxide, europium doped (Y2O3/Eu) is a luminescent material with high chemical stability and good resistance to corrosion. It is considered one of the best red oxide phosphors under the excitation wavelength (λ) of ~254 nm with a high melting point and high thermal conductivity. It can be synthesized by a variety of techniques such as sol-gel, solid-state reaction, hydrothermal synthesis, and wet chemical synthesis.[1][2][3]

Application

Y2O3/Eu can be used for a variety of sustainable applications such as dye-sensitized solar cells (DSSCs), luminescent sensors, and light-emitting displays (LEDs).[2][3][4][5]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Site-selective deposition and micropatterning of visible-light-emitting europium-doped yttrium oxide thin film on self-assembled monolayers
Masuda Y, et al.
Chemistry of Materials, 19(5), 1002-1008 (2007)
Luminescence performance of europium-doped yttrium oxide thin films
Jayaramaiah JR, et al.
Journal of Luminescence, 157(19), 63-68 (2015)
Luqing Xi et al.
Dalton transactions (Cambridge, England : 2003), 46(40), 13835-13844 (2017-10-03)
A novel red phosphor, NaHF
Structural and optical properties of europium doped yttrium oxide nanoparticles for phosphor applications
Srinivasan R, et al.
Journal of alloys and compounds, 496(1-2), 472-477 (2010)
Sensitive detection of free bilirubin in blood serum using $\beta$-diketone modified europium-doped yttrium oxide nanosheets as a luminescent sensor
Yang W, et al.
Royal Society of Chemistry Advances, 8(32), 17854-17859 (2018)

Articles

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service