Skip to Content
Merck
All Photos(1)

Key Documents

74640

Sigma-Aldrich

19-Nortestosterone

≥99.0% (HPLC)

Synonym(s):

17β-Hydroxy-19-norandrost-4-en-3-one, 17β-Hydroxy-4-estren-3-one, 19-Norandrostenolone, 4-Estren-17β-ol-3-one, Nadrolone, Nandrolone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H26O2
CAS Number:
Molecular Weight:
274.40
Beilstein:
2055849
EC Number:
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99.0% (HPLC)

form

crystals

optical activity

[α]20/D +57±2°, c = 1% in chloroform

drug control

USDEA Schedule IIIN; regulated under CDSA - not available from Sigma-Aldrich Canada

functional group

hydroxyl
ketone

SMILES string

C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@H]34)[C@@H]1CC[C@@H]2O

InChI

1S/C18H26O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h10,13-17,20H,2-9H2,1H3/t13-,14+,15+,16-,17-,18-/m0/s1

InChI key

NPAGDVCDWIYMMC-IZPLOLCNSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

19-Nortestosterone, also known as nandrolone or 17β-hydroxy-19-nor-4-androsten-3-one, is an androgenic anabolic steroid (AAS).

Application

19-Nortestosterone can be used as a reactant to prepare:
  • Fluorescent nandrolone-BODIPY conjugates to study intracellular localization and tracking of nandrolone using fluorescence microscope.
  • Optically pure hexadecahydrochrysene diastereomers, and chrysene enantiomers.

Other Notes

Sales restrictions may apply

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The use of symmetry in enantioselective synthesis: Four pairs of chrysene enantiomers prepared from 19-nortestosterone
Stastna E, et al.
Organic & Biomolecular Chemistry, 9(12), 4685-4694 (2011)
Simeon Pierre Fodouop Chegaing et al.
Steroids, 162, 108679-108679 (2020-06-23)
Although the discovery of antibiotics has decreased the spread and severity of infectious diseases, their uncontrolled use has lead to the emergence of bacterial resistance to existing chemotherapeutic agents. Bacterial disease thus remains a challenge for health authorities in worldwide
Synthesis and biological evaluation of nandrolone-bodipy conjugates
J Michal, et al.
Steroids, 97, 62-66 (2015)
Pieter Van Renterghem et al.
Analytica chimica acta, 768, 41-48 (2013-03-12)
Due to their performance enhancing properties, use of anabolic steroids (e.g. testosterone, nandrolone, etc.) is banned in elite sports. Therefore, doping control laboratories accredited by the World Anti-Doping Agency (WADA) screen among others for these prohibited substances in urine. It
P Kintz et al.
Journal of pharmaceutical and biomedical analysis, 24(5-6), 1125-1130 (2001-03-15)
A sensitive, specific and reproducible method for the quantitative determination of nandrolone in human hair has been developed. The sample preparation involved a decontamination step of the hair with methylene chloride. The hair sample (about 100 mg) was solubilized in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service