Skip to Content
Merck
All Photos(2)

Key Documents

746347

Sigma-Aldrich

Dichlorotriphenylphosphine[2-(diphenylphosphino)-N-(2-pyridinylmethyl)ethanamine]ruthenium(II)

greener alternative

97%

Synonym(s):

Ru-PNN catalyst

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H36Cl2N2P2Ru
CAS Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

reaction suitability

core: ruthenium
reagent type: catalyst

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

275-295 °C

greener alternative category

SMILES string

Cl[Ru]Cl.[H]N(CCP(C1=CC=CC=C1)C2=CC=CC=C2)CC3=NC=CC=C3.P(C4=CC=CC=C4)(C5=CC=CC=C5)C6=CC=CC=C6

InChI

1S/C20H21N2P.C18H15P.2ClH.Ru/c1-3-10-19(11-4-1)23(20-12-5-2-6-13-20)16-15-21-17-18-9-7-8-14-22-18;1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;;;/h1-14,21H,15-17H2;1-15H;2*1H;/q;;;;+2/p-2

InChI key

JFYJJNHKCBDTMZ-UHFFFAOYSA-L

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

Catalyst for hydrogenation of esters to alcohols. Greener alternative to stoichiometric hydride reagents.

From Esters to Alcohols and Back with Ruthenium and Osmium Catalysts

Legal Information

This Product is licensed from GreenCentre Canada for non-commercial, research use only. Please contact GreenCentre Canada for information regarding commercial use of Product.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Spasyuk, D.;
Angewandte Chemie (International Edition in English), 2772-2772 (2012)

Related Content

Prof. Dmitry Gusev and his co-workers are developing organometallic compounds of transition metals with tridentate ('pincer'-type) ligands for applications in catalytic hydrogenation of compounds with polar C=O and C=N bonds, dehydrogenative coupling and dehydrogenation reactions of alcohols.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service