Skip to Content
Merck
All Photos(1)

Key Documents

745774

Sigma-Aldrich

8-Amino-1-octanethiol hydrochloride

95%

Synonym(s):

8-Mercaptooctylamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H19NS · HCl
CAS Number:
Molecular Weight:
197.77
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

95%

form

solid

mp

158-163 °C

storage temp.

2-8°C

SMILES string

SCCCCCCCCN.Cl

InChI

1S/C8H19NS.ClH/c9-7-5-3-1-2-4-6-8-10;/h10H,1-9H2;1H

InChI key

CCWZBRKMJBSNQS-UHFFFAOYSA-N

General description

8-Amino-1-octanethiol hydrochloride (8-AOT) is an amine based alkanethiol that can be used as a self-assembled monolayer (SAM) on a variety of surfaces. It facilitates the immobilization of surface atoms and controls the physiochemical properties of the substrate.[1][2]

Application

8-AOT can be used in the surface functionalization of gold electrodes for the trace analysis of oligonucleotide using poly nucleic acid (PNA) based ion-channel sensors.[3] It can be used in the fabrication of peptide microarray for surface plasmon resonance based screening of protein kinase activity.[4]
Self Assembly Molecules (SAMs); the amino group is usually modified using amine-reactive materials; such as proteins or biomaterials; to functionalize the gold surface. It is used for biosenors with large amounts of immobilized protein and suppression of nonspecific adsorption

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photooxidation of amine-terminated self-assembled monolayers on gold
Lee S, et al.
The Journal of Physical Chemistry C, 114(23), 10512-10519 (2010)
Evaluation of protein kinase activities of cell lysates using peptide microarrays based on surface plasmon resonance imaging
Mori T, et al.
Analytical Biochemistry, 375(2), 223-231 (2008)
Trace analysis of an oligonucleotide with a specific sequence using PNA-based ion-channel sensors
Aoki H and Umezawa Y
Analyst, 128(6), 681-685 (2003)
Tuning the surface potential of gold substrates arbitrarily with self-assembled monolayers with mixed functional groups
Lin W, et al.
Physical Chemistry Chemical Physics, 11(29), 6199-6204 (2009)
Jolene L Lau et al.
ACS nano, 5(10), 7722-7729 (2011-09-09)
A high-affinity RNA aptamer (K(d) = 50 nM) was efficiently identified by SELEX against a heteroaryldihydropyrimidine structure, chosen as a representative drug-like molecule with no cross reactivity with mammalian or bacterial cells. This aptamer, its weaker-binding variants, and a known

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service