Skip to Content
Merck
All Photos(1)

Key Documents

734632

Sigma-Aldrich

Lanthanum strontium cobalt ferrite

greener alternative

powder, 0.6 μm particle size

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
26111700
NACRES:
NA.23

form

powder

mol wt

Mw 222.85 g/mol

composition

La0.6Sr0.4Co0.2Fe0.8O3

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

surface area

10.0-14.0 m2/g

particle size

0.40-0.80 μm (d50)
0.6 μm

bulk density

1.203 g/mL

application(s)

battery manufacturing

greener alternative category

General description

Lanthanum strontium cobalt ferrite (LSCF) belongs to the class of cathode materials that operate at low temperatures. It has mixed ionic and electronic conductivities with high electrocatalytic activity. It forms thin films with higher performance than the thick dense film.
Lanthanum strontium cobalt ferrite (LSCF) is a mixed ionic and electron conducting pervoskite. The ionic conductivity is lower than YSZ although maintaining high conductivity. X-ray diffraction and electrochemical measurements of an operational LSCF cathode was conducted to study its lattice expansion during use. LSCF can be prepared by solid state reaction of individual cation oxides.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.

Application

LSCF may be used as the cathode in solid oxide fuel cells (SOFCs).
LSCF oxygen electrodes can be used in the fabrication of high-performance solid oxide fuel cells (SOFCs).

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marina, O. A.; Pederson, L. R.; Williams, M. C.; Coffey, G. W. et al.
Journal of the Electrochemical Society, 154, B452-B452 (2007)
Low-temperature electrochemical characterization of dense ultra-thin lanthanum strontium cobalt ferrite (La0. 6Sr0. 4Co0. 8Fe0. 2O3) cathodes synthesized by RF-sputtering on nanoporous alumina-supported Y-doped zirconia membranes
Xiong H, et al.
Journal of Power Sources, 193(2), 589-592 (2009)
Development of lanthanum strontium cobalt ferrite composite cathodes for intermediate-to low-temperature solid oxide fuel cells
Baharuddin NA, et al.
Journal of Zhejiang University. Science, 14(1), 11-24 (2013)
Leone, P.; Santarelli, M.; Asinari, P.; Cali, M.; Borchiellini, R.
Journal of Power Sources, 177, 111-111 (2008)
Preparation of an asymmetric perovskite-type membrane and its oxygen permeability
Jin W, et al.
Journal of Membrane Science, 185(2), 237-243 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service