Skip to Content
Merck
All Photos(1)

Documents

733210

Sigma-Aldrich

Tetrakis(dimethylsulfoxide)dichlororuthenium(II)

96%

Synonym(s):

Dichlorotetrakis(dimethyl sulfoxide)ruthenium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H24Cl2O4RuS4
CAS Number:
Molecular Weight:
484.51
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

solid

reaction suitability

core: ruthenium
reagent type: catalyst

mp

192-198 °C

SMILES string

O=S(C)C.O=S(C)C.Cl[Ru](Cl)O=S(C)C.O=S(C)C

InChI

1S/4C2H6OS.2ClH.Ru/c4*1-4(2)3;;;/h4*1-2H3;2*1H;/q;;;;;;+2/p-2

InChI key

UMJDEUKQHKMAOI-UHFFFAOYSA-L

General description

Tetrakis(dimethylsulfoxide)dichlororuthenium(II), [RuCl2(dmso)4] is an organoruthenium compound. It is used as a homogeneous catalyst in organic synthesis.

Tetrakis(dimethylsulfoxide)dichlororuthenium(II) (Ru(DMSO)4Cl2) is a coordination complex generally used as a catalyst in various reactions. In addition, it is also used as a precursor in the synthesis of other ruthenium complexes.

Application

RuCl2(dmso)4 can be used as a catalyst for:
  • The β-alkylation of secondary alcohols via hydrogen auto-transfer process.
  • The oxidation of thioethers (S) to sulfoxides by N-methylmorpholine N-oxide (NMO).

Tetrakis(dimethylsulfoxide)dichlororuthenium(II) can be used as a precursor in the synthesis of Ru(II)-MEPE (metallo-supramolecular polyelectrolytes) complex, which is used to prepare electrochromic organic-metallic hybrid polymers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly enantioselective and cis-diastereoselective cyclopropanation of olefins catalyzed by ruthenium complexes of (iminophosphoranyl) ferrocenes
Hoang VD, et al
Tetrahedron Letters, 48(45), 8014-8017 (2007)
Electrochromic solid-state devices using organic-metallic hybrid polymers
Higuchi M, et al.
Journal of Inorganic and Organometallic Polymers and Materials, 19, 74-78 (2009)
Evolution of physical and photocatalytic properties of new Zn (II) and Ru (II) complexes
Polyhedron, 170, 412-423 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service