Skip to Content
Merck
All Photos(4)

Key Documents

730785

Sigma-Aldrich

Silver, dispersion

nanoparticles, 10 nm particle size (TEM), 0.02 mg/mL in aqueous buffer, contains sodium citrate as stabilizer

Synonym(s):

Colloidal silver

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ag
CAS Number:
Molecular Weight:
107.87
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

nanoparticles

Quality Level

contains

sodium citrate as stabilizer

concentration

0.02 mg/mL in aqueous buffer

refractive index

n20/D 1.333

particle size

10 nm (TEM)

density

0.997 g/mL at 25 °C

fluorescence

λem 388 nm FWHM 59 nm

storage temp.

2-8°C

SMILES string

[Ag]

InChI

1S/Ag

InChI key

BQCADISMDOOEFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Our silver nanoparticles (10 nm in diameter) are spherical nanoparticles of metallic silver dispersed in an aqueous buffer. The particles have high electronic conductivity, high surface area, and, because of their tightly controlled particle size, fluoresce at 388 nm with a FWHM of only 50 nm.

Application

AgNPs dispersion can be used for a variety of applications such as optoelectronics, energy harvesting, and surface enhanced Raman spectroscopy (SERS).

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Plasmonic effects of quantum size metal nanoparticles on Synthesis of highly monodisperse citrate-stabilized silver nanoparticles of up to 200 nm: kinetic control and catalytic properties
Bastus NG, et al.
Chemistry of Materials, 7(6), 2069-2083 (2017)
Synthesis and characterization of stable aqueous dispersions of silver nanoparticles through the Tollens process
Yin Y, et al.
Journal of Materials Chemistry, 12(3), 522-527 (2002)
Charlene Andraos et al.
International journal of toxicology, 39(5), 397-421 (2020-07-17)
Despite several studies addressing nanoparticle (NP) interference with conventional toxicity assay systems, it appears that researchers still rely heavily on these assays, particularly for high-throughput screening (HTS) applications in order to generate "big" data for predictive toxicity approaches. Moreover, researchers
Direct synthesis of narrowly dispersed silver nanoparticles using a single-source precursor
Lin X, et al.
Langmuir, 19(24), 10081-10085 (2003)
Direct synthesis of narrowly dispersed silver nanoparticles using a single-source precursor
Lin XZ, et al.
Langmuir, 19(24), 10081-10085 (2003)

Articles

Silver nanomaterials have unique physical, chemical, and optical properties that are currently being leveraged for a wide variety of biological applications.

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service