730114
2-Methacryloyloxyethyl phosphorylcholine
contains ≤100 ppm MEHQ as inhibitor, 97%
Synonym(s):
MPC
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
C11H22NO6P
CAS Number:
Molecular Weight:
295.27
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Quality Level
Assay
97%
form
powder or crystals
contains
≤100 ppm MEHQ as inhibitor
mp
143-148 °C
storage temp.
2-8°C
SMILES string
CC(=C)C(=O)OCCOP([O-])(=O)OCC[N+](C)(C)C
InChI
1S/C11H22NO6P/c1-10(2)11(13)16-8-9-18-19(14,15)17-7-6-12(3,4)5/h1,6-9H2,2-5H3
InChI key
ZSZRUEAFVQITHH-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
2-Methacryloyloxyethyl phosphorylcholine (MPC) is a zwitterionic phosphorylcholine group-containing methacrylate monomer and is the most suitable monomer to mimic the phospholipid polar groups contained with cell membranes. It is also used in the preparations of a wide range of polymeric biomaterials because of its excellent resistance to nonspecific protein adsorption, biocompatibility, and low toxicity. It is also used to develop diverse molecular structures with adjustable properties using a range of polymerization methods, including living radical polymerization. Due to the reactivity of methacrylate, MPC-based materials have a wide range of applications in biomedical fields. Furthermore, MPC is also used as a monomer in the production of various polymeric materials for several applications such as medical devices, lithium-based batteries, contact lenses, drug delivery, wound dressings, and tissue engineering scaffolds.
Application
2-Methacryloyloxyethyl phosphorylcholine (MPC) can be used in the following applications:
- Use as a monomer to produce fully zwitterionic polymer networks in a solvate ionic liquid for use as a gel electrolyte in lithium-based batteries.
- Synthesis of polymer coatings for medical devices to prevent blood clotting and bacterial adhesion.
- Preparation of hydrogels for use as contact lens materials and wound dressing.
- Production of polymeric micelles and nanoparticles for drug delivery.
- Use as a monomer to prepare polymer-modified antifouling silicone hydrogel contact lenses. The addition of MPC forms a cell membrane-like structure on the contact lens and helps to prevent cell and bacterial adhesion on the surface.
Features and Benefits
- It has bioinert properties due to the presence of a zwitter ionic phosphorylcholine (PC) group in the side chain.
- It can build various molecular architectures with tunable properties.
- It has excellent resistance to cell adhesion, blood coagulation, and non-specific protein adsorption.
related product
Product No.
Description
Pricing
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
N Nakabayashi et al.
Bio-medical materials and engineering, 14(4), 345-354 (2004-10-09)
Copolymers of 2-methacryloyloxyethyl phosphorylcholine (MPC) showed good hemocompatibility as hypothesized. The hypothesis was surfaces having phosphorylcholine groups by polymerization of MPC could accumulate phospholipids from blood stream and show good blood compatibility. We designed and prepared a methacylate having a
Boxuan Ma et al.
Acta biomaterialia, 70, 186-196 (2018-02-17)
Intelligent drug delivery systems with prolonged circulation time, reduced drug leakage in blood, target site-triggered drug release and endosomal escape are attractive and ideal for malignant tumor therapy. Herein, doxorubicin (DOX)-conjugated smart polymeric micelles based on 4-carboxy benzaldehyde-grafted poly (L-lysine)-block-poly
Yumiko Obayashi et al.
Frontiers in microbiology, 8, 1952-1952 (2017-10-27)
Microbial extracellular hydrolytic enzymes that degrade organic matter in aquatic ecosystems play key roles in the biogeochemical carbon cycle. To provide linkages between hydrolytic enzyme activities and genomic or metabolomic studies in aquatic environments, reliable measurements are required for many
N Nakabayashi et al.
Biomaterials, 24(13), 2431-2435 (2003-04-18)
This review addresses the non-thrombogenic characteristics of copolymers based on 2-methacryloyloxyethyl phosphorylcholine (MPC), originally developed by Nakabayashi and colleagues. The hypothesis underlying these developments was that such materials would adsorb phospholipids from blood, yielding surfaces with good natural blood compatibility.
Piers E Milner et al.
Acta biomaterialia, 65, 102-111 (2017-11-08)
Partial joint repair is a surgical procedure where an artificial material is used to replace localised chondral damage. These artificial bearing surfaces must articulate against cartilage, but current materials do not replicate both the biphasic and boundary lubrication mechanisms of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service