Skip to Content
Merck
All Photos(1)

Key Documents

727679

Sigma-Aldrich

Diethylmethyl(2-methoxyethyl)ammonium bis(trifluoromethylsulfonyl)imide

for electrochemistry, ≥98.5% (qNMR)

Synonym(s):

N,N-Diethyl-N-methyl-N-(2-methoxyethyl)ammonium bis(trifluoromethanesulfonyl)imide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20F6N2O5S2
CAS Number:
Molecular Weight:
426.40
Beilstein:
10134444
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

for electrochemistry

Assay

≥98.5% (qNMR)

form

liquid

impurities

≤500 ppm water

anion traces

bromide (Br-): ≤10 mg/kg
chloride (Cl-): ≤10 mg/kg
fluoride (F-): ≤10 mg/kg
nitrate (NO3-): ≤20 mg/kg
phosphate (PO43-): ≤10 mg/kg
sulfate (SO42-): ≤10 mg/kg

SMILES string

CC[N+](C)(CC)CCOC.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

InChI

1S/C8H20NO.C2F6NO4S2/c1-5-9(3,6-2)7-8-10-4;3-1(4,5)14(10,11)9-15(12,13)2(6,7)8/h5-8H2,1-4H3;/q+1;-1

InChI key

WUFQNPMBKMKEHN-UHFFFAOYSA-N

General description

Diethylmethyl(2-methoxyethyl)ammonium bis(trifluoromethylsulfonyl)imide is a room temperature ionic liquid (RTIL), which shows high CO2 solubility.

Application

Diethylmethyl(2-methoxyethyl)ammonium bis(trifluoromethylsulfonyl)imide can be used to develop a polymeric gel electrolyte system that shows magnesium ion (Mg2+) mobility.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Densities and low pressure solubilities of carbon dioxide in five promising ionic liquids.
Nonthanasin T, et al.
Royal Society of Chemistry Advances, 4(15), 7566-7578 (2014)
Room Temperature Electrodeposition of Metallic Magnesium from Ethylmagnesium Bromide in Tetrahydrofuran and Ionic Liquid Mixtures
Chen, Lu, et al.
Journal of the Electrochemical Society, 163.4, H3043-H3051 (2016)
A novel polymeric gel electrolyte systems containing magnesium salt with ionic liquid.
Yoshimoto N, et al.
Electrochimica Acta, 50(19), 3866-3871 (2005)
Study on the reaction of chlorophenols in room temperature ionic liquids with ionizing radiation
Kimura, Atsushi, et al.
Radiation Physics and Chemistry, 77.10, 1253-1257 (2008)
Son T Le et al.
Nanoscale, 11(33), 15622-15632 (2019-08-14)
We have demonstrated atomically thin, quantum capacitance-limited, field-effect transistors (FETs) that enable the detection of pH changes with 75-fold higher sensitivity (≈4.4 V per pH) over the Nernst value of 59 mV per pH at room temperature when used as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service