Skip to Content
Merck
All Photos(1)

Key Documents

727296

Sigma-Aldrich

(S)-1-Methoxy-2-propylamine

ChiPros®, produced by BASF, 99%

Synonym(s):

(S)-2-Amino-1-methoxypropane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H11NO
CAS Number:
Molecular Weight:
89.14
Beilstein:
5725728
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.5% (GC)
99%

form

liquid

optical purity

enantiomeric excess: ≥99.5%

functional group

amine
ether

SMILES string

COC[C@H](C)N

InChI

1S/C4H11NO/c1-4(5)3-6-2/h4H,3,5H2,1-2H3/t4-/m0/s1

InChI key

NXMXETCTWNXSFG-BYPYZUCNSA-N

Application

(S)-1-Methoxy-2-propylamine can be used:
  • As an intermediate in the synthesis of piperazinebenzylamine based human MC4 receptor antagonists.
  • To prepare imidazopyrimidine derivatives as potent p38 MAP kinase inhibitors.
  • In the synthesis of a marine natural product nhatrangin A.

Legal Information

ChiPros is a registered trademark of BASF SE

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

47.3 °F

Flash Point(C)

8.5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optimization of piperazinebenzylamines with a N-(1-methoxy-2-propyl) side chain as potent and selective antagonists of the human melanocortin-4 receptor
Pontillo J, et al.
Bioorganic & medicinal chemistry letters, 15(20), 4615-4618 (2005)
Imidazopyrimidines, potent inhibitors of p38 MAP kinase
Rupert KC, et al.
Bioorganic & Medicinal Chemistry Letters, 13(3), 347-350 (2003)
Formal enantioselective synthesis of nhatrangin A
Feuillastre S, et al.
Organic & Biomolecular Chemistry, 18(10), 1949-1956 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service