Skip to Content
Merck
All Photos(2)

Key Documents

717436

Sigma-Aldrich

Benzamide oxime

97%

Synonym(s):

N-Hydroxybenzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8N2O
CAS Number:
Molecular Weight:
136.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

74-78 °C

functional group

amine
oxime
phenyl

SMILES string

N\C(=N\O)c1ccccc1

InChI

1S/C7H8N2O/c8-7(9-10)6-4-2-1-3-5-6/h1-5,10H,(H2,8,9)

InChI key

MXOQNVMDKHLYCZ-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Clement et al.
The Journal of biological chemistry, 272(31), 19615-19620 (1997-08-01)
Drugs containing strong basic nitrogen functional groups can be N-oxygenated to genotoxic products. While the reduction of such products is of considerable toxicological significance, most in vitro studies have focused on oxygen-sensitive reductase systems. However, an oxygen-insensitive microsomal hydroxylamine reductase
A Jousserandot et al.
Biochemistry, 37(49), 17179-17191 (1998-12-23)
Oxidation by rat liver microsomes of 13 compounds involving a C=N(OH) function (including N-hydroxyguanidines, amidoximes, ketoximes, and aldoximes) was found to occur with the release of nitrogen oxides such as NO, NO2-, and NO3-. The greatest activities were observed with
A K Fröhlich et al.
Xenobiotica; the fate of foreign compounds in biological systems, 35(1), 17-25 (2005-03-25)
N-Hydroxyamidines (amidoximes) can act as pro-drugs of amidines (e.g. ximelagatran, a novel direct thrombin inhibitor). This known pro-drug principle is based on the N-reduction of an oral bioavailable amidoxime to its active form. Previous study of the metabolism of the
T L Deegan et al.
Bioorganic & medicinal chemistry letters, 9(2), 209-212 (1999-02-18)
1,2,4-Oxadiazoles have been prepared in parallel using 1,1'-carbonyldiimidazole (CDI) as a reagent for both formation and cyclodehydration of O-acyl benzamidoximes. The use of CDI facilitates parallel purification of the oxadiazole products by simple liquid-liquid extraction and filtration.
B Clement et al.
Archiv der Pharmazie, 322(7), 431-435 (1989-07-01)
At pH 7.4 neither benzamidine (1) is ring-hydroxylated nor benzamidoxime (2) is N-hydroxylated, reduced or ring-hydroxylated by aerobic incubations with microsomal fractions (12000 g supernatant, microsomes) of rabbit liver homogenates and NADPH. Products of hydrolytic processes are also not detected.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service