Skip to Content
Merck
All Photos(3)

Key Documents

716820

Sigma-Aldrich

Gold nanorods

10 nm diameter, λmax, 808 nm, dispersion in H2O

Synonym(s):

Au nanorods, Gold nanorod

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

colloidal suspension
dispersion in H2O
nanorod

Quality Level

contains

CTAB as stabilizer

concentration

>30 μg/mL in H2O

diam. × L

10 nm × 41 nm ± 10%

impurities

<0.1% CTAB
<0.1% ascorbic acid

diameter

10 nm

pH

3-5

density

1 g/mL at 25 °C

λmax

808 nm

Mw/Mn

(<10% CV, monodispersity)
>95 (rods)

storage temp.

2-8°C

SMILES string

[Au]

InChI

1S/Au

InChI key

PCHJSUWPFVWCPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Gold nanorods (AuNRs) are gold nanoparticles that have been elongated to form a rod type structure. They are interesting materials due to their optical properties that allow a strong absorption band in the visible part of the spectrum. They can easily be tuned to achieve spectra in a wide range of wavelength.

Application

AuNRs are mainly utilized in the biological application as materials that are suitable in sensors, photothermal therapy and other imaging based devices.

Physical properties

Do not freeze, quality lost if frozen even once. Please keep at approximately 4 °C

Legal Information

This product was produced under the methods claimed in U.S. Pat. Nos. 8,956,440 and 8,241,922

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bioproduction of gold nanoparticles for photothermal therapy
Silva CO, et al.
Therapeutic Delivery, 7(5), 287-304 (2016)
Biological applications of gold nanorods
Stone J, et al.
Wiley Interdisciplinary Reviews. Nanomedicine and Nanobiotechnology, 3(1), 100-109 (2011)
Gold nanorods: synthesis, characterization and applications
Perez-Juste J, et al.
Coordination Chemistry Reviews, 249(17-18), 1870-1901 (2005)
Light interactions with gold nanorods and cells: implications for photothermal nanotherapeutics
Ungureanu C, et al.
Nano Letters, 11(5), 1887-1894 (2011)

Articles

Gold nanostructures such as nanorods, nanowires and microgold have found applications in exciting fields such as biomedical engineering, catalysis and diagnostics.

Gold nanostructures such as nanorods, nanowires and microgold have found applications in exciting fields such as biomedical engineering, catalysis and diagnostics.

Gold nanostructures such as nanorods, nanowires and microgold have found applications in exciting fields such as biomedical engineering, catalysis and diagnostics.

Gold nanostructures such as nanorods, nanowires and microgold have found applications in exciting fields such as biomedical engineering, catalysis and diagnostics.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service