Skip to Content
Merck
All Photos(1)

Key Documents

709255

Sigma-Aldrich

2-Bromobenzyl alcohol-(phenyl-13C6)

99 atom % 13C, 98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br13C6H4CH2OH
CAS Number:
Molecular Weight:
192.99
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

98% (CP)

form

solid

mp

78-80 °C (lit.)

mass shift

M+6

SMILES string

OC[13c]1[13cH][13cH][13cH][13cH][13c]1Br

InChI

1S/C7H7BrO/c8-7-4-2-1-3-6(7)5-9/h1-4,9H,5H2/i1+1,2+1,3+1,4+1,6+1,7+1

InChI key

IOWGHQGLUMEZKG-ZFJHNFROSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service