Skip to Content
Merck
All Photos(1)

Documents

705594

Sigma-Aldrich

Cortisol-9,11,12,12-d4

≥98 atom % D, ≥98% (CP)

Synonym(s):

11β,17α,21-Trihydroxypregn-4-ene-3,20-dione-D4, 11β,17α,21-Trihydroxypregn-4-ene-3,20-dione-d4 (9,11,12,12-d4), 17-Hydroxycorticosterone-9,11,12,12-d4, 17-Hydroxycorticosterone-D4, 4-Pregnene-11β,17α,21-triol-3,20-dione-D4, 4-Pregnene-11β,17α,21-triol-3,20-dione-d4 (9,11,12,12-d4), Deuterated cortisol, Deuterated hydrocortisone, Hydrocortisone-9,11,12,12-d4, Kendall′s compound F (9,11,12,12-d4), Reichstein′s substance M (9,11,12,12-d4)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21D4H26O5
CAS Number:
Molecular Weight:
366.48
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

isotopic purity

≥98 atom % D

Quality Level

Assay

≥98% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

mp

211-214 °C

mass shift

M+4

storage temp.

−20°C

SMILES string

[2H]C1([2H])[C@]([2H])(O)[C@@]2([2H])[C@@H](CCC3=CC(=O)CC[C@]23C)[C@@H]4CCC(O)(C(=O)CO)[C@@]14C

InChI

1S/C21H30O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h9,14-16,18,22,24,26H,3-8,10-11H2,1-2H3/t14-,15-,16-,18+,19-,20-,21-/m0/s1/i10D2,16D,18D

InChI key

JYGXADMDTFJGBT-BJJKTIQHSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Therina du Toit et al.
The Journal of steroid biochemistry and molecular biology, 166, 54-67 (2016-06-28)
Adrenal C
Mayako Saito-Abe et al.
Pediatrics international : official journal of the Japan Pediatric Society, 62(7), 785-788 (2020-01-25)
Sampling of salivary cortisol is non-invasive and important for the evaluation of the hypothalamic-pituitary-adrenal axis function and stress levels. However, the reference values for salivary cortisol measured by liquid chromatography-tandem mass spectrometry (LC-MS/MS) in healthy infants are unclear. The aim
Lillian R Sturmer et al.
Steroids, 129, 35-40 (2017-12-05)
Late night salivary cortisol measurement is a clinically important and convenient screening test for Cushing's syndrome. Tandem mass spectrometry (LC-MS/MS) assays have superior sensitivity and specificity compared to immunoassays. Our goal was to improve a LC-MS/MS method to measure salivary
Susanna Kortesluoma et al.
Psychoneuroendocrinology, 124, 105064-105064 (2020-12-02)
Previous research suggests that maternal prenatal psychological distress (PPD) is related to altered cortisol reactivity in the exposed child. There are indications for the sex differences in vulnerability for prenatal adversities that depend on the exposure and child outcome. Still
Sergi Coll et al.
Drug testing and analysis, 13(2), 348-359 (2020-09-20)
Betamethasone (BET) is prohibited in sports competitions when administered by systemic routes, and it is allowed by other routes for therapeutic purposes. In out-of-competition periods, there is no restriction of use. The present work aimed to assess the urinary excretion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service