Skip to Content
Merck
All Photos(1)

Key Documents

704393

Sigma-Aldrich

Lithium tantalate

≥99.99%

Synonym(s):

Lithium tantalum trioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiTaO3
CAS Number:
Molecular Weight:
235.89
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

Assay

≥99.99%

form

powder

impurities

≤100.0 ppm Trace Metal Analysis

mp

>300 °C

SMILES string

[Li+].[O-][Ta](=O)=O

InChI

1S/Li.3O.Ta/q+1;;;-1;

InChI key

CIFJATMCNLSYQG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maidul Beg et al.
Journal of biomolecular structure & dynamics, 37(13), 3536-3549 (2018-09-04)
Nanomaterials in different form have been thoroughly used in the area of pharmaceutics and medicine for drug delivery. The large scale of nanoparticles (NPs) synthesis from plant extract is much safe, cheap and eco-friendly. Here, we demonstrated a new, one-step
Gautam Patel et al.
Journal of nanoscience and nanotechnology, 19(8), 5054-5073 (2019-03-28)
Two-dimensional (2D) materials have rewritten many disciplines of modern science and technology, both through intensive fundamental research. In this review, we will provide a comprehensive summary of the properties, synthesis, characterization, and application of 2D materials (Graphene, Hexagonal boron nitride
Mousumi Saha et al.
The journal of physical chemistry. A, 124(23), 4690-4699 (2020-05-13)
In this study, atomic level interactions of a 1:1 choline chloride (ChCl)/acetylsalicylic acid (ASA) therapeutic deep eutectic solvent (THDES) has been investigated by combining the molecular dynamics (MD), density functional theory (DFT), and spectroscopic (Raman and IR) techniques. Atom-atom radial
Valmala Bhavana et al.
Talanta, 199, 679-688 (2019-04-07)
Niclosamide, an anthelmintic drug recently repurposed for its activity against cancer, crystallizes into three solvated forms, two monohydrates (NHa, NHb) and one anhydrous (NAn) form. NAn is sensitive to pseudopolymorphic transformations that affect its dissolution and consequently, its bioavailability. NAn
Alexandra Muñoz-Bonilla et al.
International journal of molecular sciences, 19(12) (2018-12-24)
Antimicrobial polymers and coatings are potent types of materials for fighting microbial infections, and as such, they have attracted increased attention in many fields. Here, a series of antimicrobial copolymers were prepared by radical copolymerization of 2-hydroxyethyl methacrylate (HEMA), which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service