Skip to Content
Merck
All Photos(2)

Key Documents

695580

Sigma-Aldrich

1,3-Butadiene solution

20 wt. % in toluene

Synonym(s):

Bivinyl, Vinylethylene, alpha,gamma-Butadiene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6
CAS Number:
Molecular Weight:
54.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

liquid

concentration

20 wt. % in toluene

refractive index

n20/D 1.493

density

0.806 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

C=CC=C
C=CC=C

InChI

1S/C4H6/c1-3-4-2/h3-4H,1-2H2
1S/C4H6/c1-3-4-2/h3-4H,1-2H2

InChI key

KAKZBPTYRLMSJV-UHFFFAOYSA-N
KAKZBPTYRLMSJV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,3-Butadiene can be used:
  • In the synthesis of 1,6-hexanedial (adipic aldehyde) by bis-hydroformylation using rhodium as a catalyst.
  • As a monomer for the synthesis of cis-1,4-polybutadiene by polymerization using cobalt(II) and nickel(II) complex catalysts.
  • As a precursor for the selective synthesis of butene by hydrogenation reaction.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 1A - Flam. Liq. 2 - Muta. 1B - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-105.0 °F

Flash Point(C)

-76.11 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toward the Rhodium-Catalyzed Bis-Hydroformylation of 1, 3-Butadiene to Adipic Aldehyde
Smith SE, et al.
Organometallics, 30(13), 3643-3651 (2011)
Single-atom Pd1/graphene catalyst achieved by atomic layer deposition: remarkable performance in selective hydrogenation of 1, 3-butadiene
Yan H, et al.
Journal of the American Chemical Society, 137(33), 10484-10487 (2015)
Polymerization of 1, 3-butadiene catalyzed by cobalt (II) and nickel (II) complexes bearing imino-or amino-pyridyl alcohol ligands in combination with ethylaluminum sesquichloride
Ai Pengfei, et al.
Journal of Organometallic Chemistry, 705(13), 51-58 (2012)
Masato Ohashi et al.
Journal of the American Chemical Society, 133(45), 18018-18021 (2011-10-19)
Pyridines, which comprise one of the most important classes of the six-membered heterocyclic compounds, are widely distributed in nature, and the transition-metal-catalyzed [2 + 2 + 2] cycloaddition reaction of two alkynes and a nitrile is one of the most
Matthew S McCammant et al.
Journal of the American Chemical Society, 135(11), 4167-4170 (2013-03-12)
A palladium-catalyzed 1,4-addition across the commodity chemical 1,3-butadiene to afford skipped polyene products is reported. Through a palladium σ → π → σ allyl isomerization, two new carbon-carbon bonds are formed with high regioselectivity and trans stereoselectivity of the newly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service