Skip to Content
Merck
All Photos(2)

Key Documents

693839

Sigma-Aldrich

6-Phosphonohexanoic acid

97%

Synonym(s):

PHA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13O5P
CAS Number:
Molecular Weight:
196.14
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

97%

form

solid

mp

98-104 °C

SMILES string

OC(=O)CCCCCP(O)(O)=O

InChI

1S/C6H13O5P/c7-6(8)4-2-1-3-5-12(9,10)11/h1-5H2,(H,7,8)(H2,9,10,11)

InChI key

SYFTUNQVCGSBOV-UHFFFAOYSA-N

General description

6-Phosphonohexanoic acid (PHA) is an alkylphosphonic acid that forms a self-assembled monolayer (SAM) on a variety of surfaces. It has carboxylic groups that facilitate the grafting of the surface atoms while the phosphonic groups allow the surface modification of the substrate material.[1]

Application

PHA can be used in the surface functionalization of phynox material which can further be utilized in the biomedical applications such as surgical implant and stents.[2] It can also be used in dispersing manganese dioxide (MnO2)-MWCNT for the formation of electrodes for supercapacitors.[3] It may also be coated on indium tin oxide (ITO) as an alternative to silane based SAMs for the fabrication of organic electronic devices such as organic solar cells (OSCs).[4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Surface modification of alumina-coated silica nanoparticles in aqueous sols with phosphonic acids and impact on nanoparticle interactions
Pauly CS, et al.
Physical Chemistry Chemical Physics, 17(29), 19173-19182 (2015)
Elena Herrera-Carrillo et al.
PloS one, 12(5), e0177935-e0177935 (2017-05-26)
Allogeneic transplantation of blood stem cells from a CCR5-Δ32 homozygous donor to an HIV-infected individual, the "Berlin patient", led to a cure. Since then there has been a search for approaches that mimic this intervention in a gene therapy setting.
Mariana E Mendes et al.
International journal of radiation biology, 95(8), 1058-1071 (2019-04-23)
Purpose: The goal was to compare the micronucleus (MN) and dicentric plus ring chromosomes (D + R) assays for dose assessment in cases of partial body irradiations (PBI). Materials and methods: We constructed calibration curves for each assay at doses ranging from
Channarena Chuon et al.
Scientific reports, 9(1), 12186-12186 (2019-08-23)
Approximately 75% of hepatocellular carcinomas (HCC) occur in Asia; core promoter mutations are associated with HCC in HBV genotype C, the dominant genotype in Cambodia. We analyzed these mutations in Cambodian residents and compared them with HBV full genomes registered
Hee Taek Kim et al.
Polymers, 11(7) (2019-07-25)
Cadaverine is a C5 diamine monomer used for the production of bio-based polyamide 510. Cadaverine is produced by the decarboxylation of l-lysine using a lysine decarboxylase (LDC). In this study, we developed recombinant Escherichia coli strains for the expression of

Articles

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service