Skip to Content
Merck
All Photos(1)

Key Documents

68351

Sigma-Aldrich

Trifluoracetaldehyde monohydrate

technical, ~75% in H2O

Synonym(s):

2,2,2-Trifluoro-1,1-ethanediol, Fluoral hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2HF3O · H2O
CAS Number:
Molecular Weight:
116.04
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

form

liquid

concentration

~75% in H2O

refractive index

n20/D 1.340

bp

104-106 °C (lit.)

density

1.44 g/mL at 20 °C (lit.)

functional group

aldehyde

SMILES string

O.FC(F)(F)C=O

InChI

1S/C2HF3O.H2O/c3-2(4,5)1-6;/h1H;1H2

InChI key

INDFRFZGABIRRW-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-9.4 °F

Flash Point(C)

-23.0 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service