Skip to Content
Merck
All Photos(1)

Key Documents

67260

Sigma-Aldrich

5-Methyl-3-heptanone

≥94% (GC)

Synonym(s):

Ethyl isoamyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH(CH3)CH2COCH2CH3
CAS Number:
Molecular Weight:
128.21
Beilstein:
506345
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥94% (GC)

refractive index

n20/D 1.414 (lit.)

bp

157-162 °C (lit.)

density

0.823 g/mL at 25 °C (lit.)

functional group

ketone

SMILES string

CCC(C)CC(=O)CC

InChI

1S/C8H16O/c1-4-7(3)6-8(9)5-2/h7H,4-6H2,1-3H3

InChI key

PSBKJPTZCVYXSD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • 5-Methyl-3-heptanone can be converted into the corresponding gem-dihydroperoxide using aqueous hydrogen peroxide and chlorosulfonic acid as a catalyst at room temperature.
  • It can be reduced to its corresponding alcohol using sodium borohydride in urea/choline chloride eutectic salt.
  • It can undergo condensation with tert-butyl-sulfinamide to form the corresponding chiral tert-butyl-sulfinyl-ketimine, which can further react with ylides derived from trimethylsulfonium iodide and S-allyl tetrahydrothiophenium bromide to form highly substituted chiral aziridines.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

111.2 °F - closed cup

Flash Point(C)

44 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An efficient synthesis of gem-dihydroperoxides and 1, 2, 4, 5-tetraoxanes catalyzed bychlorosulfonic acid as a new catalyst.?
Khosravi K and Asgari A
JOURNAL OF ADVANCES IN CHEMISTRY, 11(3), 3381-3390 (2015)
Natural deep eutectic salt promoted regioselective reduction of epoxides and carbonyl compounds.
Azizi N, et al.
Royal Society of Chemistry Advances, 2(6), 2289-2293 (2012)
Direct synthesis of chiral aziridines from N-tert-butyl-sulfinylketimines.
Morton D, et al.
Chemical Communications (Cambridge, England), (17), 1833-1835 (2006)
Denis Schenkel et al.
Frontiers in microbiology, 9, 1847-1847 (2018-08-29)
The plant-growth modulating effect of microbial volatile organic compounds (VOCs) has been demonstrated repeatedly. This has most often been performed by exposing plants to VOC released by microbes grown on nutrient rich media. Here, we used soil instead to grow

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service