Skip to Content
Merck
All Photos(2)

Key Documents

667846

Sigma-Aldrich

BBL

Synonym(s):

Poly(benzimidazobenzophenanthroline)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C20H6N4O2)n
CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

description

Band gap: 1.9 eV

Quality Level

form

solid

Orbital energy

HOMO -5.9 eV 
LUMO -4.0 eV 

OPV Device Performance

ITO/MEH-PPV/BBL/Al

  • Short-circuit current density (Jsc): 1.98 mA/cm2
  • Open-circuit voltage (Voc): 0.93 V
  • Fill Factor (FF): 0.47
  • Power Conversion Efficiency (PCE): 1.1 %

ITO/PPV/BBL/Al
  • Short-circuit current density (Jsc): 2.15 mA/cm2
  • Open-circuit voltage (Voc): 1.1 V
  • Fill Factor (FF): 0.5
  • Power Conversion Efficiency (PCE): 1.5 %

semiconductor properties

N-type (mobility=0.1 cm2/V·s)
P-type (mobility=0.4 cm2/V·s)

Application

Polymer semiconductor with n-type behavior in organic field-effect transistors (FETs) and photovoltaic cells (PVs). Processed from solutions in methanesulfonic acid.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jenekhe, S.A.; Yi, S.
Applied Physics Letters, 77, 2635-2635 (2000)
Efficient Solar Cells from Layered Nanostructures of Donor and Acceptor Conjugated Polymers
Alam, M. M.; Jenekhe, S. A.
Chemistry of Materials, 16, 4647-4656 (2004)
Manoj, A.G.; Narajan, K.S.
Opt. Mater., 21, 417-417 (2002)
Amit Babel et al.
Journal of the American Chemical Society, 125(45), 13656-13657 (2003-11-06)
Field-effect mobility of electrons as high as 0.1 cm2/(V s) is observed in n-channel thin film transistors fabricated from a solution spin-coated conjugated ladder polymer, poly(benzobisimidazobenzophenanthroline) (BBL), under ambient air conditions. This is the highest electron mobility observed to date

Articles

The development of high-performance conjugated organic molecules and polymers has received widespread attention in industrial and academic research.

Intrinsically stretchable active layers for organic field-effect transistors (OFET) are discussed. Polymer structural modification & post-polymerization modifications are 2 methods to achieve this.

Fabrication procedure of organic field effect transistor device using a soluble pentacene precursor.

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service