All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H10F3N
CAS Number:
Molecular Weight:
153.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
refractive index
n20/D 1.393
bp
74-75 °C/33 mmHg
density
1.154 g/mL at 25 °C
functional group
fluoro
storage temp.
2-8°C
SMILES string
FC(F)(F)C1CCCNC1
InChI
1S/C6H10F3N/c7-6(8,9)5-2-1-3-10-4-5/h5,10H,1-4H2
InChI key
JOHFJTBDUSVGQB-UHFFFAOYSA-N
Related Categories
Application
Reactant for synthesis of positive allosteric modulators of metabotropic glutamate receptor subtype 4
Reactant for 5-HT7 receptor ligands
Reactant for 5-HT7 receptor ligands
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
82.4 °F - closed cup
Flash Point(C)
28 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service