Skip to Content
Merck
All Photos(3)

Key Documents

665363

Sigma-Aldrich

(S,R,R)-(+)-(3,5-Dioxa-4-phosphacyclohepta[2,1-a:3,4-a′]dinaphthalen-4-yl)bis(1-phenylethyl)amine

95% (HPLC)

Synonym(s):

(+)-N,N-Bis[(1R)-1-phenylethyl]-dinaphtho[2,1-d:1′,2′-f][1,3,2]dioxaphosphepin-4-amine, (11bS)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H30NO2P
CAS Number:
Molecular Weight:
539.60
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95% (HPLC)

mp

102-103 °C

functional group

amine
phenyl

SMILES string

C[C@@H](N([C@H](C)c1ccccc1)P2Oc3ccc4ccccc4c3-c5c(O2)ccc6ccccc56)c7ccccc7

InChI

1S/C36H30NO2P/c1-25(27-13-5-3-6-14-27)37(26(2)28-15-7-4-8-16-28)40-38-33-23-21-29-17-9-11-19-31(29)35(33)36-32-20-12-10-18-30(32)22-24-34(36)39-40/h3-26H,1-2H3/t25-,26-/m1/s1

InChI key

LKZPDRCMCSBQFN-CLJLJLNGSA-N

Application

Chiral phosphoramidite ligand used in a highly enantioselective copper-catalyzed conjugate addition of diakylzincs to a variety of Michael acceptors. Also used in a palladium-catalyzed diethylzinc mediated umpolung allylation.

Features and Benefits

Advantages of the MonoPhos® ligands:
  • Superior enantiocontrol in numerous transformations
  • High activities at low catalyst loadings
  • Hydrogenations under low-pressure conditions

Legal Information

Sold under license from DSM for research purposes only.
MonoPhos is a registered trademark of DSM IP Assets B.V.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oliver Knopff et al.
Organic letters, 4(22), 3835-3837 (2003-02-26)
[formula: see text] The zinc enolates, resulting from the copper-catalyzed enantioselective conjugate addition of dialkyl zinc reagents to cyclic and acyclic enones, could be trapped, quantitatively, as silyl enol ethers with TMSOTf in apolar solvents or with TMSCI and NEt3.
Privileged Ligands
Aldrich Chemfiles, 6(8), 7-7 null
Organic & Biomolecular Chemistry, 4, 1278-1278 (2006)
Synlett, 1375-1375 (2001)
Jorge Esquivias et al.
The Journal of organic chemistry, 70(18), 7451-7454 (2005-08-27)
[reaction: see text] The enantioselective catalytic 1,4-addition to alpha,beta-unsaturated ketimines is an unprecedented process. Herein, we document the copper-catalyzed addition of dialkylzinc reagents to (2-pyridylsulfonyl)imines of chalcones. This process occurs rapidly in the presence of a chiral phosphoramidite ligand to

Articles

DSM collaboration offers MonoPhos™ ligands for research, based on the BINOL platform by Feringa and co-workers.

DSM collaboration offers MonoPhos™ ligands for research, based on the BINOL platform by Feringa and co-workers.

DSM collaboration offers MonoPhos™ ligands for research, based on the BINOL platform by Feringa and co-workers.

DSM collaboration offers MonoPhos™ ligands for research, based on the BINOL platform by Feringa and co-workers.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service